Merck
  • Simultaneous determination of 15 nitroimidazoles in cosmetics by HPLC coupled with electrospray ionization- tandem mass spectrometry.
Simultaneous determination of 15 nitroimidazoles in cosmetics by HPLC coupled with electrospray ionization- tandem mass spectrometry.
Journal of AOAC International (2015-01-30)
Xian-Shuang Meng, Hua Bai, Qing Zhang, Qing Lv, Yun-Xia Chen, Hui-Juan Ma, Jing-Rui Li, Qiang Ma
ABSTRACT
A sensitive and reliable analytical method based on HPLC/MSIMS has been developed for the simultaneous determination of 15 nitroimidazoles in cosmetics. A diversity of cosmetic samples, including powder, lotion, shampoo, and cream were collected. The samples were ultrasonically extracted with aqueous methanol, and the extracts were then subjected to cleanup bySPE using an Oasis HLB cartridge followed by filtration with a 0.20 pm membrane filter. Afterwards, chromatographic separation was performed on an XSelect CSH C18 column (2.1 x 150 mm, 3.5 pm) maintained at 30°C within 15 min by a gradient of acetonitrile-0.1% aqueous formic acid solution at a flow rate of 0.25 mL/min. The mass spectrometric detection was carried, out using electrospray positive ionization under the multiple reaction monitoring mode. A good linearity was observed over the concentration range from 0.5 to 500 ng/mL. The intraday and interday precisions, which were investigated by determining all target compounds in cosmetics seven times/day and on 7 consecutive days, were below 5.00%. The mean recoveries at three spiked levels ranged from 80.42 to 100.83% with the RSDs from 0.45 to 9.02%. The LOQs were determined to be between 0.01 and 0.1 mg/kg. The method was sufficiently rapid, reliable, and sensitive for the determination of 15 nitroimidazoles in cosmetics.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Ronidazole, analytical standard
Supelco
Tinidazole, VETRANAL®, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Tinidazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Ipronidazole, VETRANAL®, analytical standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, analytical standard
Supelco
Ipronidazole-OH, VETRANAL®, analytical standard