Merck
  • Modest hyperglycemia prevents interstitial dispersion of insulin in skeletal muscle.
Modest hyperglycemia prevents interstitial dispersion of insulin in skeletal muscle.
Metabolism: clinical and experimental (2014-12-04)
Cathryn M Kolka, Ana Valeria B Castro, Erlinda L Kirkman, Richard N Bergman
ABSTRACT
Insulin injected directly into skeletal muscle diffuses rapidly through the interstitial space to cause glucose uptake, but this is blocked in insulin resistance. As glucotoxicity is associated with endothelial dysfunction, the observed hyperglycemia in diet-induced obese dogs may inhibit insulin access to muscle cells, and exacerbate insulin resistance. Here we asked whether interstitial insulin diffusion is reduced in modest hyperglycemia, similar to that induced by a high fat diet. During normoglycemic (100 mg/dl) and moderately hyperglycemic (120 mg/dl) clamps in anesthetized canines, sequential doses of insulin were injected into the vastus medialis of one hindlimb; the contra-lateral limb served as a control. Plasma samples were collected and analyzed for insulin content. Lymph vessels of the hind leg were also catheterized, and lymph samples were analyzed as an indicator of interstitial insulin concentration. Insulin injection increased lymph insulin in normoglycemic animals, but not in hyperglycemic animals. Muscle glucose uptake was elevated in response to hyperglycemia, however the insulin-mediated glucose uptake in normoglycemic controls was not observed in hyperglycemia. Modest hyperglycemia prevented intra-muscularly injected insulin from diffusing through the interstitial space reduced insulin-mediated glucose uptake. Hyperglycemia prevents the appearance of injected insulin in the interstitial space, thus reducing insulin action on skeletal muscle cells.
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
USP
Acepromazine maleate, United States Pharmacopeia (USP) Reference Standard
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acepromazine maleate, ≥98% (HPLC)
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder