Merck
  • Comparison of the therapeutic effect between sodium bicarbonate and insulin on acute propafenone toxicity.
Comparison of the therapeutic effect between sodium bicarbonate and insulin on acute propafenone toxicity.
The American journal of emergency medicine (2014-08-26)
Hwa Yeon Yi, Jang Young Lee, Won Suk Lee, Won Young Sung, Sang Won Seo
ABSTRACT
Unlike other sodium-channel-blocking antiarrhythmic agents, propafenone has β-blocking effects and calcium-channel-blocking effects. Yi et al recently studied insulin's treatment effect on acute propafenone toxicity in rats. However, because the degree of effectiveness of insulin compared to the previously known antidote sodium bicarbonate (NaHCO3) was not studied, the 2 treatment methods were compared for propafenone intoxication in rats. Rats received intravenous propafenone (36 mg/[kg h]) for 12 minutes. After the induction of toxicity, rats (n = 10 per group) received normal saline solution (NSS), NaHCO3, or insulin with glucose as treatment. Animals in the NSS, NaHCO3, and Insulin groups received an intravenous infusion of 36 mg/(kg h) propafenone until death occurred. For each animal, the mean arterial pressure (MAP, heart rate, PR interval, QRS duration, total hemoglobin, sodium, potassium, potential of hydrogen, bicarbonate, glucose, lactate, and central venous oxygen saturation (Scvo2) were measured and compared among the groups. Survival of the Insulin group was greater than that of the NSS group by log-rank test (P = .021). Sodium bicarbonate prevented the decline of MAP for 55 minutes. In comparison, insulin prevented the decline of MAP and heart rate, and the elongation of the PR interval and QRS duration for 55 minutes (P < .05). Propafenone toxicity led to decreased Ca(2+), potential of hydrogen, and Scvo2 and increased lactate levels. Insulin prevented the decrease of Ca(2+) and Scvo2, whereas NaHCO3 prevented the increase in lactate. Insulin treatment was more effective than NaHCO3 on acute propafenone toxicity in rat. Therefore, when propafenone-induced cardiotoxicity occurs, which is unresponsive to current treatment methods, glucose-insulin infusion may be considered.
Product Description
Propafenone hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Propafenone hydrochloride
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Lactic acid, natural, ≥85%
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Sodium, 25-35 wt % dispersion in paraffin
Sigma-Aldrich
Sodium, ACS reagent, dry
Sigma-Aldrich
Sodium, cubes, contains mineral oil, 99.9% trace metals basis
Sigma-Aldrich
Sodium, 99.95% trace metals basis, ingot
Sigma-Aldrich
Sodium, in kerosene, pieces (large), ≥99.8% (sodium basis)
Sigma-Aldrich
DL-Lactic acid, ~90% (T)
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%