Merck
  • Inhibition of the mitochondrial pyrimidine biosynthesis enzyme dihydroorotate dehydrogenase by doxorubicin and brequinar sensitizes cancer cells to TRAIL-induced apoptosis.
Inhibition of the mitochondrial pyrimidine biosynthesis enzyme dihydroorotate dehydrogenase by doxorubicin and brequinar sensitizes cancer cells to TRAIL-induced apoptosis.
Oncogene (2013-09-10)
T He, S Haapa-Paananen, V O Kaminskyy, P Kohonen, V Fey, B Zhivotovsky, O Kallioniemi, M Perälä
ABSTRACT
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) is a promising agent in selectively killing tumor cells. However, TRAIL monotherapy has not been successful as many cancer cells are resistant to TRAIL. Chemotherapeutic agents, such as doxorubicin have been shown to act synergistically with TRAIL, but the exact mechanisms of actions are poorly understood. In this study, we performed high-throughput small interfering RNA screening and genome-wide gene expression profiling on doxorubicin-treated U1690 cells to explore novel mechanisms underlying doxorubicin-TRAIL synergy. The screening and expression profiling results were integrated and dihydroorotate dehydrogenase (DHODH) was identified as a potential candidate. DHODH is the rate-limiting enzyme in the pyrimidine synthesis pathway, and its expression was downregulated by doxorubicin. We demonstrated that silencing of DHODH or inhibition of DHODH activity by brequinar dramatically increased the sensitivity of U1690 cells to TRAIL-induced apoptosis both in 2D and 3D cultures, and was accompanied by downregulation of c-FLIPL as well as by mitochondrial depolarization. In addition, uridine, an end product of the pyrimidine synthesis pathway was able to rescue the sensitization effects initiated by both brequinar and doxorubicin. Furthermore, several other cancer cell lines, LNCaP, MCF-7 and HT-29 were also shown to be sensitized to TRAIL by brequinar. Taken together, our findings have identified a novel protein target and its inhibitor, brequinar, as a potential agent in TRAIL-based combinatorial cancer therapy and highlighted for the first time the importance of mitochondrial DHODH enzyme and pyrimidine pathway in mediating TRAIL sensitization in cancer cells.
Sigma-Aldrich
Pyrimidine, ≥98.0%
Sigma-Aldrich
Monoclonal Anti-PARP antibody produced in mouse, clone C-2-10, ascites fluid
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
TRAIL human, recombinant, expressed in NSO cells, >97% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
TRAIL human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE and HPLC), lyophilized powder
Sigma-Aldrich
MISSION® esiRNA, targeting human DHODH
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
USP
Leflunomide, United States Pharmacopeia (USP) Reference Standard
Leflunomide for peak identification, European Pharmacopoeia (EP) Reference Standard
Leflunomide, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Dhodh
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source