Merck
  • The design and synthesis of alanine-rich α-helical peptides constrained by an S,S-tetrazine photochemical trigger: a fragment union approach.
The design and synthesis of alanine-rich α-helical peptides constrained by an S,S-tetrazine photochemical trigger: a fragment union approach.
The Journal of organic chemistry (2013-12-24)
Joel R Courter, Mohannad Abdo, Stephen P Brown, Matthew J Tucker, Robin M Hochstrasser, Amos B Smith
ABSTRACT
The design and synthesis of alanine-rich α-helical peptides constrained in a partially unfolded state by incorporation of the S,S-tetrazine phototrigger has been achieved, permitting, upon photochemical release, observation by 2D-IR spectroscopy of the subnanosecond conformational dynamics that govern the early steps associated with α-helix formation. Solid-phase peptide synthesis was employed to elaborate the requisite fragments, with full peptide construction via solution-phase fragment condensation. The fragment union tactic was also employed to construct (13)C═(18)O isotopically edited amides to permit direct observation of conformational motion at or near specific peptide bonds.
Sigma-Aldrich
3,6-Dichloro-1,2,4,5-tetrazine, 96%