Skip to Content
Merck
All Photos(2)

Documents

792594

Sigma-Aldrich

3,6-Dichloro-1,2,4,5-tetrazine

96%

Synonym(s):

Dichloro-s-tetrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2Cl2N4
Molecular Weight:
150.95
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

availability

available only in USA

mp

146-151 °C

storage temp.

2-8°C

SMILES string

ClC1=NN=C(Cl)N=N1

InChI

1S/C2Cl2N4/c3-1-5-7-2(4)8-6-1

InChI key

HXXFMIAFWKZHDY-UHFFFAOYSA-N

Application

Reagent has been reported in stapling of complex peptides as a photochemical trigger. Professor Amos Smith and coworkers have displayed this application in several recent reports.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Self-react. C - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen P Brown et al.
Journal of the American Chemical Society, 137(12), 4034-4037 (2015-03-21)
Protocols have been achieved that permit facile introduction of s-tetrazine into unprotected peptides and the protein, thioredoxin, between two cysteine sulfhydryl groups (i.e., staple), followed by photochemical release (i.e., unstaple) and regeneration of the peptide/protein upon removal of the cyano
Joel R Courter et al.
The Journal of organic chemistry, 79(2), 759-768 (2013-12-24)
The design and synthesis of alanine-rich α-helical peptides constrained in a partially unfolded state by incorporation of the S,S-tetrazine phototrigger has been achieved, permitting, upon photochemical release, observation by 2D-IR spectroscopy of the subnanosecond conformational dynamics that govern the early
Mohannad Abdo et al.
Organic letters, 14(13), 3518-3521 (2012-06-27)
The design, solid-phase synthesis, and photochemical validation of diverse peptide linchpins, containing the S,S-tetrazine phototrigger, have been achieved. Steady state irradiation or femtosecond laser pulses confirm their rapid photofragmentation. Attachment of peptides to the C- and N-termini will provide access
Matthew J Tucker et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(43), 17314-17319 (2013-10-10)
The relaxation of helical structures very close to equilibrium is observed via transient 2D IR spectroscopy. An initial distribution of synthetically distorted helices having an unnatural bridge linking the 10th and 12th residues of an alanine-rich α-helix is released to
Tetrazine phototriggers: probes for peptide dynamics.
Matthew J Tucker et al.
Angewandte Chemie (International ed. in English), 49(21), 3612-3616 (2010-04-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service