Merck
Camphor toxicity.
Pediatric clinics of North America (1986-04-01)
E Siegel, S Wason
ABSTRACT
Camphor is present in several over-the-counter compounds of questionable use and therefore may be ingested by small children. Because seizures may follow ingestion of certain amounts, appropriate treatment is needed, including the use of anticonvulsants.
Sigma-Aldrich
(1R)-(+)-Camphor, 98%
Supelco
(−)-Camphor, analytical standard
USP
Camphor, United States Pharmacopeia (USP) Reference Standard
Supelco
D-Camphor, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(1S)-(−)-Camphor, 95%
Supelco
D-Camphor, analytical standard
Sigma-Aldrich
D-Camphor, ≥97%, FG