Skip to Content
Merck
All Photos(1)

Key Documents

21293

Supelco

(−)-Camphor

analytical standard

Synonym(s):

(1S)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
1907612
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

optical activity

[α]20/D −43±1°, c = 10% in ethanol

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

CC1(C)[C@H]2CC[C@]1(C)C(=O)C2

InChI

1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m0/s1

InChI key

DSSYKIVIOFKYAU-OIBJUYFYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey N Love et al.
The Journal of emergency medicine, 27(1), 49-54 (2004-06-29)
Serious pediatric toxicity resulting from exposure to small amounts of camphor-containing products has long been a problem. Twenty years ago the United States Food and Drug Administration took several actions in an attempt to ameliorate this risk. Despite these changes
Camphor: a chiral starting material in natural product synthesis.
T Money
Natural product reports, 2(3), 253-289 (1985-06-01)
Hiroyuki Kitahata et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 87(1), 010901-010901 (2013-02-16)
The coupling between deformation and motion in a self-propelled system has attracted broader interest. In the present study, we consider an elliptic camphor particle for investigating the effect of particle shape on spontaneous motion. It is concluded that the symmetric
Christian Merten et al.
The journal of physical chemistry. A, 116(27), 7329-7336 (2012-06-06)
In the present work, the first observation of strong resonance Raman optical activity (RROA) involving more that one resonant electronic state is reported. The chiral transition metal complex bis-(trifluoroacetylcamphorato) copper(II), abbreviated Cu(tfc)(2), exhibits both resonance Raman (RR) and RROA spectra
Yiji Lin et al.
Dalton transactions (Cambridge, England : 2003), 41(22), 6696-6706 (2012-04-28)
Chiral tetrakis(β-diketonate) Ln(III) complexes Δ-[NaLa(d-hfc)(4)(CH(3)CN)] (1) and Λ-[NaLa(l-hfc)(4) (CH(3)CN)] (2) (d/l-hfc(-) = 3-heptafluo-robutylryl-(+)/(-)-camphorate) are a pair of enantiomers and crystallize in the same Sohncke space group (P2(1)2(1)2(1)) with dodecahedral (DD) geometry. Typically positive and negative exciton splitting patterns around 320

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service