- Sulfadiazine-induced nephrolithiasis in children.
Sulfadiazine-induced nephrolithiasis in children.
Pediatric nephrology (Berlin, Germany) (2004-06-19)
Charlotte Catalano-Pons, Sophie Bargy, Deborah Schlecht, Marie-Dominique Tabone, George Deschênes, Albert Bensman, Tim Ulinski
PMID15206032
ABSTRACT
Sulfadiazine-associated urinary calculi have been described in HIV-positive adult patients but rarely in children. We report two pediatric cases of sulfadiazine-induced nephrolithiasis and review 45 adult cases from the literature. One had a hyper-IgM syndrome and was treated with sulfadiazine for cerebral toxoplasmosis, the other had toxoplasmic retinitis. Both developed multiple bilateral lithiasis with acute renal failure. Normalization of renal function and reduction of calculi size was rapidly achieved after discontinuation of sulfadiazine, hyperhydration, and alkalinization. Bilateral ureteral stents were required in one patient because of anuria.
Product Description
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
Sulfadiazine for identification of impurity F, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sulfadiazine, European Pharmacopoeia (EP) Reference Standard