Merck
  • High LET (56)Fe ion irradiation induces tissue-specific changes in DNA methylation in the mouse.
High LET (56)Fe ion irradiation induces tissue-specific changes in DNA methylation in the mouse.
Environmental and molecular mutagenesis (2014-04-12)
Florence Lima, Dacheng Ding, Wilfried Goetz, Austin J Yang, Janet E Baulch
ABSTRACT
DNA methylation is an epigenetic mechanism that drives phenotype and that can be altered by environmental exposures including radiation. The majority of human radiation exposures occur in a relatively low dose range; however, the biological response to low dose radiation is poorly understood. Based on previous observations, we hypothesized that in vivo changes in DNA methylation would be observed in mice following exposure to doses of high linear energy transfer (LET) (56) Fe ion radiation between 10 and 100 cGy. We evaluated the DNA methylation status of genes for which expression can be regulated by methylation and that play significant roles in radiation responses or carcinogenic processes including apoptosis, metastasis, cell cycle regulation, and DNA repair (DAPK1, EVL, 14.3.3, p16, MGMT, and IGFBP3). We also evaluated DNA methylation of repeat elements in the genome that are typically highly methylated. No changes in liver DNA methylation were observed. Although no change in DNA methylation was observed for the repeat elements in the lungs of these same mice, significant changes were observed for the genes of interest as a direct effect and a delayed effect of irradiation 1, 7, 30, and 120 days post exposure. At delayed times, differences in methylation profiles among genes were observed. DNA methylation profiles also significantly differed based on dose, with the lowest dose frequently affecting the largest change. The results of this study are the first to demonstrate in vivo high LET radiation-induced changes in DNA methylation that are tissue and locus specific, and dose and time dependent.
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, foil, 25mm disks, thickness 0.015mm, 99.85%
Iron, foil, 15mm disks, thickness 0.1mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.01mm, 99.99+%
Iron, foil, 20m coil, thickness 0.038mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.05mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.05mm, as rolled, 99.99+%
Iron, foil, 50mm disks, thickness 0.038mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.9mm, as rolled, 99.5%
Iron, foil, 25mm disks, thickness 0.125mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.05mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.38mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.025mm, as rolled, 99.99+%
Iron, foil, 50mm disks, thickness 0.075mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.20mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.5mm, hard, 99.5%
Iron, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.998%
Iron, foil, 8mm disks, thickness 0.125mm, hard, 99.5%
Iron, foil, 8mm disks, thickness 0.05mm, hard, 99.5%
Iron, foil, light tested, 100x100mm, thickness 0.025mm, hard, 99.5%
Iron, rod, 1000mm, diameter 2.0mm, as drawn, 99.99+%
Iron, foil, not light tested, 100x100mm, thickness 0.007mm, 99.85%
Iron, foil, light tested, 100x100mm, thickness 0.038mm, hard, 99.5%
Iron, foil, not light tested, 100x100mm, thickness 0.003mm, 99.85%
Iron, rod, 100mm, diameter 25mm, as drawn, 98+%