Skip to Content
Merck
All Photos(3)

Documents

267945

Sigma-Aldrich

Iron

chips, 99.98% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
EC Number:
MDL number:
UNSPSC Code:
12141721
PubChem Substance ID:
NACRES:
NA.23

Assay

99.98% trace metals basis

form

chips

resistivity

9.71 μΩ-cm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Iron fortification and malaria risk in children.
Andrew M Prentice et al.
JAMA, 310(9), 914-915 (2013-09-05)
Stanley Zlotkin et al.
JAMA, 310(9), 938-947 (2013-09-05)
In sub-Saharan Africa, malaria is a leading cause of childhood morbidity and iron deficiency is among the most prevalent nutritional deficiencies. In 2006, the World Health Organization and the United Nations Children's Fund released a joint statement that recommended limiting
Shaun D Wong et al.
Nature, 499(7458), 320-323 (2013-07-23)
Mononuclear non-haem iron (NHFe) enzymes catalyse a broad range of oxidative reactions, including halogenation, hydroxylation, ring closure, desaturation and aromatic ring cleavage reactions. They are involved in a number of biological processes, including phenylalanine metabolism, the production of neurotransmitters, the
N M Makarova et al.
Klinicheskaia laboratornaia diagnostika, (1)(1), 42-44 (2013-07-03)
The sample for examination included 52 patients with chronic cardiac failure and 26 patients with anemia amongst. The expected increase of production of prohepsidin against the background of increased serum concentrations of anti-inflammatory cytokines was not detected. The tendency to
Tomasz Pytrus et al.
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego, 34(203), 263-268 (2013-07-31)
The aim of the study was to analyze the occurrence of anemia in children with newly diagnosed Crohn's disease and 3 months after the initiation of treatment depending on location and activity of the disease. 36 children aged from 2

Articles

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service