Merck
  • Mitochondrial metabolism and stress response of yeast: Applications in fermentation technologies.
Mitochondrial metabolism and stress response of yeast: Applications in fermentation technologies.
Journal of bioscience and bioengineering (2013-11-12)
Hiroshi Kitagaki, Hiroshi Takagi
ABSTRACT
Mitochondria are sites of oxidative respiration. During sake brewing, sake yeasts are exposed to long periods of hypoxia; the structure, role, and metabolism of mitochondria of sake yeasts have not been studied in detail. It was first elucidated that the mitochondrial structure of sake yeast transforms from filamentous to dotted structure during sake brewing, which affects malate metabolism. Based on the information of yeast mitochondria during sake brewing, practical technologies have been developed; (i) breeding pyruvate-underproducing sake yeast by the isolation of a mutant resistant to an inhibitor of mitochondrial pyruvate transport; and (ii) modifying malate and succinate production by manipulating mitochondrial activity. During the bread-making process, baker's yeast cells are exposed to a variety of baking-associated stresses, such as freeze-thaw, air-drying, and high sucrose concentrations. These treatments induce oxidative stress generating reactive oxygen species due to mitochondrial damage. A novel metabolism of proline and arginine catalyzed by N-acetyltransferase Mpr1 in the mitochondria eventually leads to synthesis of nitric oxide, which confers oxidative stress tolerance on yeast cells. The enhancement of proline and arginine metabolism could be promising for breeding novel baker's yeast strains that are tolerant to multiple baking-associated stresses. These new and practical methods provide approaches to improve the processes in the field of industrial fermentation technologies.
Product Description
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
Proline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Proline, 99%, FCC, FG
SAFC
L-Proline
Sigma-Aldrich
L-(−)-Malic acid disodium salt, ≥95% (titration)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
USP
L-Proline, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Supelco
D-Malic acid, analytical standard
Sigma-Aldrich
D-(+)-Malic acid, unnatural form, ≥97.0% (T)
Sigma-Aldrich
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
Sigma-Aldrich
Succinic acid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Succinic acid, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Succinic acid, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
DL-Malic acid, ≥98% (capillary GC)
Supelco
DL-Malic acid, analytical standard
Sigma-Aldrich
Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T)
Sigma-Aldrich
DL-Malic acid, ReagentPlus®, ≥99%
Supelco
Succinic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Succinic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Malic acid, 99%
Supelco
Malic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Malic acid, meets USP/NF testing specifications