MilliporeSigma
  • Identification of 3-hydroxyanthranilic acid in mixed lymphocyte cultures.
Identification of 3-hydroxyanthranilic acid in mixed lymphocyte cultures.
Biological chemistry Hoppe-Seyler (1985-01-01)
E R Werner, H Lutz, D Fuchs, A Hausen, C Huber, D Niederwieser, W Pfleiderer, G Reibnegger, J Troppmair, H Wachter
ABSTRACT
3-Hydroxyanthranilic acid was identified in lymphocyte cultures during allogeneic stimulation. The metabolite and the synthetic product have identical HPLC retention times, fluorescence spectra, fluorescence intensities as a function of pH value and electrochemical behaviour.
Sigma-Aldrich
3-Hydroxyanthranilic acid, 97%