Merck
  • Phosphomolybdic and phosphotungstic acids as efficient catalysts for the synthesis of bridged 1,2,4,5-tetraoxanes from β-diketones and hydrogen peroxide.
Phosphomolybdic and phosphotungstic acids as efficient catalysts for the synthesis of bridged 1,2,4,5-tetraoxanes from β-diketones and hydrogen peroxide.
Organic & biomolecular chemistry (2013-03-01)
Alexander O Terent'ev, Ivan A Yaremenko, Vera A Vil', Igor K Moiseev, Sergey A Kon'kov, Valery M Dembitsky, Dmitri O Levitsky, Gennady I Nikishin
ABSTRACT
Phosphomolybdic acid (PMA) and phosphotungstic acid (PTA) efficiently catalyze the addition of H2O2 to β-diketones to form bridged 1,2,4,5-tetraoxanes. These reactions are not accompanied by the formation of monocyclic peroxides containing hydroxy and hydroperoxide groups or polymeric peroxides. The use of these catalysts made it possible to obtain bridged tetraoxanes from easily oxidizable benzoylacetone derivatives and α-unsubstituted β-diketones. The syntheses are scaled up to ten grams. The resulting peroxides can be easily isolated from the reaction mixture by column chromatography. The yield of tetraoxanes depends on the structure of β-diketone and varies from 12 to 83%. NMR monitoring of two bridged 1,2,4,5-tetraoxanes synthesis was carried out.
Sigma-Aldrich
Molybdenum, wire, diam. 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Phosphomolybdic acid solution, spray reagent, 10% in ethanol
Sigma-Aldrich
Phosphomolybdic Acid Solution
Sigma-Aldrich
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Sodium phosphomolybdate hydrate, technical
Sigma-Aldrich
Phosphomolybdic acid solution, 20 wt. % in ethanol
Sigma-Aldrich
Phosphomolybdic acid solution, ready-to-use spray and plunge reagent, for chromatography
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis