Skip to Content
Merck
All Photos(2)

Key Documents

203823

Sigma-Aldrich

Molybdenum

powder, <150 μm, 99.99% trace metals basis

Synonym(s):

Molybdenum element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
EC Number:
MDL number:
UNSPSC Code:
12141727
eCl@ss:
38180801
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder

resistivity

5.0 μΩ-cm, 20°C

particle size

<150 μm

bp

4612 °C (lit.)

mp

2617 °C (lit.)

density

10.3 g/mL at 25 °C (lit.)

SMILES string

[Mo]

InChI

1S/Mo

InChI key

ZOKXTWBITQBERF-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joyee Mitra et al.
Inorganic chemistry, 52(6), 3032-3042 (2013-03-07)
A series of [Mo(IV)O(mnt)(SR)(N-N)](-) (mnt = maleonitriledithiolate; R = Ph, nap, p-Cl-Ph, p-CO2H-Ph, and p-NO2-Ph; N-N = 2,2'-bipyridine (bipy) and 1,10-phenanthroline (phen)) complexes analogous to the reduced active site of enzymes of the sulfite oxidase family has been synthesized and
Phillip Ringel et al.
The Journal of biological chemistry, 288(20), 14657-14671 (2013-03-30)
Nitrate reductase (NR) is a complex molybdenum cofactor (Moco)-dependent homodimeric metalloenzyme that is vitally important for autotrophic organism as it catalyzes the first and rate-limiting step of nitrate assimilation. Beside Moco, eukaryotic NR also binds FAD and heme as additional
Tortelier, J.; Gougeon, P.
Inorganic Chemistry, 37, 6229-6229 (1998)
Nuclear nonproliferation. Iranian 'medical' reactor stokes search for alternatives.
Richard Stone
Science (New York, N.Y.), 340(6139), 1392-1392 (2013-06-22)
Copper ameliorates fluoride toxicity in fluoride and molybdenum fed rabbits.
Arjun L Khandare et al.
Biomedical and environmental sciences : BES, 26(4), 311-313 (2013-03-29)

Articles

Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.

Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service