Merck
  • Effect of ferrous sulfate fortification in germinated brown rice on seed iron concentration and bioavailability.
Effect of ferrous sulfate fortification in germinated brown rice on seed iron concentration and bioavailability.
Food chemistry (2013-02-16)
Yanyan Wei, M J I Shohag, Feng Ying, Xiaoe Yang, Chunyong Wu, Yuyan Wang
ABSTRACT
The present study evaluated the effectiveness of germination and iron fortification on iron concentration and bioavailability of brown rice. Iron fortification during germination process with 0.05-2 g/L ferrous sulfate increased the iron concentration in germinated brown rice from 1.1 to 15.6 times than those in raw brown rice. Based on the recommended dietary allowance of iron, maximum germination rate and γ-aminobutyric acid, we recommend the brown rice fortified with 0.25 g/L FeSO(4) as a suitable fortification level to use in germination process. Iron fortification during the germination process has a positive effect on iron concentration and bioavailability. A significant difference was observed among the cultivars in respect to the capacity for iron accumulation and bioavailability. Germination alone could improve in vitro iron solubility, but had no effect on iron bioavailability in Caco-2 cell, the additional fortification process should be combined to get high amount of bioavailable iron from the brown rice.
Sigma-Aldrich
Iron(II) sulfate heptahydrate, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-104.5% (manganometric)