Merck
  • Plasmodium falciparum antigen 332 is a resident peripheral membrane protein of Maurer's clefts.
Plasmodium falciparum antigen 332 is a resident peripheral membrane protein of Maurer's clefts.
PloS one (2012-11-28)
Sandra Nilsson, Davide Angeletti, Mats Wahlgren, Qijun Chen, Kirsten Moll
ABSTRACT
During the intraerythrocytic development of Plasmodium falciparum, the malaria parasite remodels the host cell cytosol by inducing membranous structures termed Maurer's clefts and inserting parasite proteins into the red blood cell cytoskeleton and plasma membrane. Pf332 is the largest known asexual malaria antigen that is exported into the red blood cell cytosol where it associates with Maurer's clefts. In the current work, we have utilized a set of different biochemical assays to analyze the solubility of the endogenous Pf332 molecule during its trafficking from the endoplasmic reticulum within the parasite to the host cell cytosol. Solubilization studies demonstrate that Pf332 is synthesized and trafficked within the parasite as a peripheral membrane protein, which after export into the host cell cytosol associates with the cytoplasmic side of Maurer's clefts in a peripheral manner. By immunofluorescence microscopy and flow cytometry, we show that Pf332 persists in close association with Maurer's clefts throughout trophozoite maturation and schizogony, and does not become exposed at the host cell surface. Our data also indicate that Pf332 interacts with the host cell cytoskeleton, but only in very mature parasite stages. Thus, the present study describes Pf332 as a resident peripheral membrane protein of Maurer's clefts and suggests that the antigen participates in host cytoskeleton modifications at completion of the intraerythrocytic developmental cycle.
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate decahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate decahydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate monohydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, 99.5-100.5% (ex dried subst.)
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate monohydrate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate monohydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent