Merck
  • The effects of QuikClot Combat Gauze on hemorrhage control in the presence of hemodilution.
The effects of QuikClot Combat Gauze on hemorrhage control in the presence of hemodilution.
U.S. Army Medical Department journal (2012-09-26)
Don Johnson, Samantha Agee, Amanda Reed, Brian Gegel, James Burgert, John Gasko, Michael Loughren
ABSTRACT
Although hemostatic agents may be effective at stopping hemorrhage, they may fail because of hemodilution from intravenous fluids. The purpose of this study was to investigate the effects of QuikClot Combat Gauze (QCG) on rebleeding in a class II hemorrhage in the presence of hemodilution in a lethal femoral injury. This was a prospective experimental, between swine subjects design. Pigs were assigned to one of two groups: QCG (n=15) or control (n=15). Thirty percent of the pig's blood was exsanguinated and then a 3:1 ratio of ringers lactate was administered. A groin injury was created by transecting the femoral artery and vein to simulate a battlefield injury and allowed to bleed for one minute. After one minute of hemorrhage, proximal pressure was applied to the injury, and QCG was placed into the wound followed by standard wound packing. The control group underwent the same procedures with the exception of the hemostatic agent. For both groups, 5 minutes of direct pressure was applied to the wound followed by a standard pressure dressing. Dressings were removed after 30 minutes, and the amount of hemorrhage was calculated in milliliters for each group for a period of 5 minutes. An activated clotting time was used to exclude any pigs with coagulation pathology. A multivariate analysis of variance indicated that there were no significant differences in the groups relative to weight, amount of one minute hemorrhage, fluid deficit replacement, blood volume, and the activated clotting time (P>.05) indicating that the groups were equivalent on these parameters. A t test indicated that there was significantly less bleeding (P=.002) in the QCG group (36 mL±112 mL) compared to the control group (340 mL±297 ml). QCG produces a robust clot that can more effectively tolerate hemodilution compared to a control group.
Sigma-Aldrich
Halloysite nanoclay
Sigma-Aldrich
Kaolin
Sigma-Aldrich
Kaolin, tested according to Ph. Eur.
Sigma-Aldrich
Kaolin, heavy, powder
Sigma-Aldrich
Kaolin, meets USP testing specifications