Skip to Content
Merck
All Photos(4)

Key Documents

685445

Sigma-Aldrich

Halloysite nanoclay

Synonym(s):

Kaolin clay

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2Si2O5(OH)4 · 2 H2O
CAS Number:
Molecular Weight:
294.19
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
nanopowder

grade

for analytical purposes

form

nanopowder

diam. × L

30-70 nm × 1-3 μm , nanotube

surface area

64 m2/g

color

75-96, Hunter Brightness

refractive index

n20/D 1.54

pore size

1.26-1.34 mL/g pore volume

capacity

8.0 meq/g cation exchange capacity

density

2.53 (true specific gravity)

SMILES string

O.O.O[Si](O)(O[Al]=O)O[Si](O)(O)O[Al]=O

InChI

1S/2Al.H4O7Si2.2H2O.2O/c;;1-8(2,3)7-9(4,5)6;;;;/h;;1-2,4-5H;2*1H2;;/q2*+1;-2;;;;

InChI key

HEFIDAICLOZRRO-UHFFFAOYSA-N

General description

Halloysite nanoclay has a tube like morphology. It shows low electrical, thermal conductivity and strong hydrogen interactions, on account of which the inner hydroxyl groups show greater stability than the surface hydroxyl groups in halloysite. The loss of hole water occurs at nearly 300 °C.
Similar to montmorillonite, halloysite consists of two layers of aluminosilicate in 1:1 Al:Si ratio. Significant improvements in thermal, mechanical properties have been reported on addition of halloysite nanotubes in polymer composites.

Application

Halloysite is frequently used as a reinforcement in various nanocomposites, some of which are listed here:
Isotactic propylene-halloysite nanocomposites.
Alloy composites by electroless/ autocatalytic deposition.
Poly(butylene terephthalate) (PBT) composites.
Zinc–halloysite nanotube (HNT) composite coatings.
Halloysite nanotube–hydroxyapatite–hyaluronic acid films.
Halloysite combined with polyamide, polypropylene, poly(ε-caprolactone) and fluroelastomers.
Its tube like morphology, high aspect ratio, and low percolation enables interesting applications as reinforcements in nanocomposites.

Legal Information

Product of Applied Minerals, Inc. Customers may obtain additional technical support and purchase commercial quantities at appliedminerals.com

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrophoretic deposition of composite halloysite nanotube?hydroxyapatite?hyaluronic acid films
Deen I and Zhitomirsky I
J. Alloy Compounds, 586(1), S531-S534 (2014)
Effects of filler type on the nonisothermal crystallization kinetics of poly(butylene terephthalate) (PBT) composites
Oburo?lu N, et al.
Journal of Applied Polymer Science, 123(1), 77-91 (2012)
Raman spectroscopy of isotactic polypropylene-halloysite nanocomposites
Ibrahim EE, et al.
Journal of Nanomaterials, 12 (2012)
Wagner A. et al.
Industrial Biotechnology (New Rochelle, N.Y.), 1, 190-190 (2005)
Du M. et al.
Eur. Polymer J., 42, 1362-1362 (2006)

Articles

Halloysite Nanotubes in Nanomaterials Research. Halloysite, a naturally occurring aluminosilicate nanotube, has been undeservedly forgotten.

Nanoclays like montmorillonite and bentonite are used in polymer-clay nanocomposites, rheology modifiers, and drug delivery carriers.

Nanoclays like montmorillonite and bentonite are used in polymer-clay nanocomposites, rheology modifiers, and drug delivery carriers.

Nanoclays like montmorillonite and bentonite are used in polymer-clay nanocomposites, rheology modifiers, and drug delivery carriers.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service