Merck
  • Enhancement of surface graft density of MPEG on alginate/chitosan hydrogel microcapsules for protein repellency.
Enhancement of surface graft density of MPEG on alginate/chitosan hydrogel microcapsules for protein repellency.
Langmuir : the ACS journal of surfaces and colloids (2012-08-28)
Jiani Zheng, Hongguo Xie, Weiting Yu, Mingqian Tan, Faquan Gong, Xiudong Liu, Feng Wang, Guojun Lv, Wanfa Liu, Guoshuang Zheng, Yan Yang, Weiyang Xie, Xiaojun Ma
ABSTRACT
Alginate/chitosan/alginate (ACA) hydrogel microcapsules were modified with methoxy poly(ethylene glycol) (MPEG) to improve protein repellency and biocompatibility. Increased MPEG surface graft density (n(S)) on hydrogel microcapsules was achieved by controlling the grafting parameters including the buffer layer substrate, membrane thickness, and grafting method. X-ray photoelectron spectroscopy (XPS) model was employed to quantitatively analyze n(S) on this three-dimensional (3D) hydrogel network structure. Our results indicated that neutralizing with alginate, increasing membrane thickness, and in situ covalent grafting could increase n(S) effectively. ACAC(PEG) was more promising than ACC(PEG) in protein repellency because alginate supplied more -COO(-) negative binding sites and prevented MPEG from diffusing. The n(S) increased with membrane thickness, showing better protein repellency. Moreover, the in situ covalent grafting provided an effective way to enhance n(S), and 1.00 ± 0.03 chains/nm(2) was achieved, exhibiting almost complete immunity to protein adsorption. This antifouling hydrogel biomaterial is expected to be useful in transplantation in vivo.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 20,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 10,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 5,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 550
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn ~2,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 750
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, BioUltra, 500
Sigma-Aldrich
Methoxypolyethylene glycol 350, average mol wt 350