Merck
Connexin-based channels in astrocytes: how to study their properties.
Methods in molecular biology (Clifton, N.J.) (2011-12-07)
Christian Giaume, Juan A Orellana, Verónica Abudara, Juan C Sáez
ABSTRACT
A typical feature of astrocytes is their high level of connexin expression. These membrane proteins constitute the molecular basis of two types of channels: gap junction channels that allow direct cytoplasm-to-cytoplasm communication and hemichannels that provide a pathway for exchanges between the intra- and extracellular media. An unusual property of these channels is their permeability for ions but also for small signaling molecules. They support intercellular communication that contribute to dynamic neuroglial interaction and interplay with neuronal activity and survival. Here, we describe multiple techniques based either on electrophysiological approaches or the monitoring of dye intercellular diffusion and uptake that permits an investigation of the properties of gap junction channels and hemichannels, respectively. These techniques are applied in astrocyte studies using in vitro models, mainly primary cultures and acute brain slices.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES sodium salt solution, BioReagent, 1M, suitable for cell culture
Sigma-Aldrich
HEPES sodium salt, ≥99.0% (titration)
Sigma-Aldrich
HEPES sodium salt, BioPerformance Certified, suitable for cell culture, ≥99.0%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
SAFC
HEPES
SAFC
HEPES sodium salt
Sigma-Aldrich
HEPES sodium salt, ≥99.5% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
HEPES solution, BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture