Skip to Content
Merck
All Photos(4)

Key Documents

H4034

Sigma-Aldrich

HEPES

HEPES

BioPerformance Certified, ≥99.5% (titration), suitable for cell culture

Synonym(s):

4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O4S
CAS Number:
Molecular Weight:
238.30
Beilstein:
883043
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

grade

BioPerformance Certified

Quality Level

Assay

≥99.5% (titration)

form

crystalline powder

storage condition

dry at room temperature

technique(s)

cell culture | mammalian: suitable
competitive inhibition ELISA: suitable

impurities

endotoxin and total aerobic microbial count, tested

color

white

useful pH range

6.8-8.2

pKa (25 °C)

7.5

cation traces

Fe: ≤5 ppm

absorption

≤0.05 at 260 in H2O at 0.1 M
≤0.05 at 280 in H2O at 0.1 M

suitability

suitable for Western blot
suitable for component for culture media

application(s)

clinical research
diagnostic assay manufacturing
general analytical
life science and biopharma

foreign activity

DNase, RNase, NICKase, protease, none detected

SMILES string

OCCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

HEPES buffer does not confer cytotoxic effects on cells and thus can be used in animal cell cultures.[1]
HEPES has been described as one of the best all-purpose buffers available for biological research.[2] At biological pH, the molecule is zwitterionic, and is effective as a buffer at pH 6.8 to 8.2. HEPES has been used in a wide variety of applications, including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.[3]
HEPES, or 4-(2-hydroxyethyl)piperazine-1-ethane-sulfonic acid, is a zwitterionic N-substituted aminosulfonic acid buffer. It demonstrates outstanding buffering capacity in the pH range of 6.8 to 8.2, boasting a pKa of 7.48 at 25°C. Widely acknowledged as one of the most versatile buffers, HEPES is extensively utilized in cell biology, biochemical, and biological research. A notable attribute is its non-reactivity with metal ions. Unlike many buffers, HEPES does not form significant complexes with most metals, making it suitable for applications involving metal ions without affecting their activity. This characteristic enhances its utility as a "non-coordinating buffer." In cell culture, HEPES excels in maintaining a stable physiological pH, even amidst fluctuations in carbon dioxide concentration resulting from cellular respiration. This sets it apart from bicarbonate buffers (NaHCO3), which, though commonly used, are less effective in pH stability.

HEPES also proves advantageous in various biological and biochemical processes. Its ampholytic nature makes it suitable as a separator for creating pH gradients in isoelectric focusing, a technique often employed in protein separation and analysis. Moreover, HEPES exhibits minimal interference with DNA-restriction enzyme reactions compared to buffers with fewer substituted amine groups, such as Tris. This makes it a preferred choice for applications involving DNA manipulation and analysis. Beyond these specific applications, HEPES is valuable in numerous other biological and biochemical processes, including immunoprecipitation, cell lysis, and live cell imaging. Its versatility, coupled with exceptional pH buffering capacity and minimal interaction with other molecules, establishes HEPES as an indispensable tool across diverse research domains.

Application

HEPES has been used:
  • To supplement Dulbecco′s modified Eagle′s medium to culture and maintain cell lines[4][5]
  • As a component of platelet suspension buffer[6]
  • To supplement Hank′s basic salt solution, which is used to wash pancreatic tissue[7]
  • As a component of wash buffer and blocking buffer in the purification and quantification of protein with enzyme-linked immunosorbent (ELISA) assay[8]
  • For the adjustment and maintenance of pH of biological solutions[8]
  • As a component of Hank′s balanced salt solution (HBSS) and dissociation medium to study neuronal development[9]
  • For homogenization of tissue and in the preparation of cytosolic and nuclear extract from cells[10]
  • As a component of keratinocyte and fibroblast culture medium[11]

Features and Benefits

  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications
  • Highly soluble in water with a useful pH range of 6.8 - 8.2 and pKa of 7.5 at 25 °C
  • Negligible metal ion binding
  • Less toxic to cells than other buffers such as Tris and phosphate
  • Stable in a wide pH range
  • Tested for Endotoxins and Total Aerobic Microbial Count
  • Free from DNase, NICKase, RNase, Endonuclease, Exonuclease and Protease

Packaging

The DRYPOUR® packaging system for high-quality hygroscopic salts DRYPOUR® packaging:
  • The current DRYPOUR® packaging configuration is in a 50L nestable drum
  • 25KG-DP are the standard prepack offering
  • Smaller volumes can be put in the same drum for variable configurations
The benefits of DRYPOUR®:
  • Improves operational efficiency by reducing material preparation time and increasing operator safety
  • Minimizes caking of hygroscopic salts
  • Offers triple protection: against moisture inside and outside, and against contamination
How does the DRYPOUR® packaging system Improve your operational efficiency?
  • This state-of-the-art design packaging system minimizes caking, thus significantly reducing the time needed for material preparation
  • PE drum with our new tamper-evident seal, a polyethylene liner with integrated desiccant bags and a breathable interior Tyvek®liner
  • Desiccant bags are firmly integrated into the polyethylene liner
  • Triple protection delivers two invaluable results:
  1. Dramatically reduced caking
  2. No contamination risk from the desiccant

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

DRYPOUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Tyvek is a registered trademark of E. I. du Pont de Nemours and Company

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Actin filament elasticity and retrograde flow shape the force-velocity relation of motile cells.
Zimmermann J, et al.
Biophysical Journal, 102(2), 287-295 (2012)
Use of a new buffer in the culture of animal cells.
Williamson JD and Cox P
The Journal of General Virology, 2, 309-309 (1968)
Complex thermorheology of living cells.
Schmidt BUS, et al.
New Journal of Physics, 17(7), 073010-073010 (2015)
Airway epithelial cell PPAR? modulates cigarette smoke-induced chemokine expression and emphysema susceptibility in mice.
Solleti SK et al.
American Journal of Physiology. Lung Cellular and Molecular Physiology, 309, L293-L293 (2015)
The slice overlay assay: a versatile tool to study the influence of extracellular signals on neuronal development.
Polleux F and Ghosh A
Science's STKE : Signal Transduction Knowledge Environment, 136, pl9-pl9 (2002)

Articles

Ensure optimal cell culture outcomes with ultrapure, endotoxin-free water. Learn about the impact of water impurities on mammalian cell cultures and how to achieve optimal cell growth.

Protocols

Formaldehyde cross-linking protocol detects specific protein interactions with confidence.

Formaldehyde cross-linking protocol detects specific protein interactions with confidence.

Formaldehyde cross-linking protocol detects specific protein interactions with confidence.

Formaldehyde cross-linking protocol detects specific protein interactions with confidence.

See All

Related Content

Find solutions for bulk production, custom packaging, and personalized chemical services to meet your unique research, development, and manufacturing requirements in your industries, including pharmaceuticals, biotechnology, and research institutions.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. Chicago, IL
    • Review 1
    • Votes 4
    5 out of 5 stars.

    Great product

    This is a classical Sigma HEPS powder that I have been using since my PhD research in the graduate school for the last 15 years.

    Helpful?

  2. 2 Ratings-Only Reviews

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service