Merck
Engineering microbial biofuel tolerance and export using efflux pumps.
Molecular systems biology (2011-05-11)
Mary J Dunlop, Zain Y Dossani, Heather L Szmidt, Hou Cheng Chu, Taek Soon Lee, Jay D Keasling, Masood Z Hadi, Aindrila Mukhopadhyay
ABSTRACT
Many compounds being considered as candidates for advanced biofuels are toxic to microorganisms. This introduces an undesirable trade-off when engineering metabolic pathways for biofuel production because the engineered microbes must balance production against survival. Cellular export systems, such as efflux pumps, provide a direct mechanism for reducing biofuel toxicity. To identify novel biofuel pumps, we used bioinformatics to generate a list of all efflux pumps from sequenced bacterial genomes and prioritized a subset of targets for cloning. The resulting library of 43 pumps was heterologously expressed in Escherichia coli, where we tested it against seven representative biofuels. By using a competitive growth assay, we efficiently distinguished pumps that improved survival. For two of the fuels (n-butanol and isopentanol), none of the pumps improved tolerance. For all other fuels, we identified pumps that restored growth in the presence of biofuel. We then tested a beneficial pump directly in a production strain and demonstrated that it improved biofuel yields. Our findings introduce new tools for engineering production strains and utilize the increasingly large database of sequenced genomes.
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
Sigma-Aldrich
3-Methyl-1-butanol, reagent grade, 98%
Sigma-Aldrich
3-Methyl-1-butanol, ACS reagent, ≥98.5%
Supelco
3-Methylbutanol, analytical standard
Sigma-Aldrich
Isoamyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
3-Methylbutanol, BioReagent, ≥98.5%
Sigma-Aldrich
3-Methylbutanol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Isoamyl alcohol, ≥98%, FG