Skip to Content
Merck
All Photos(1)

Documents

59092

Supelco

3-Methylbutanol

analytical standard

Synonym(s):

3-Methyl-1-butanol, Isoamyl alcohol, Isopentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1718835
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.8% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.1% 2-methylbutanol (remarks in GC)

refractive index

n20/D 1.407

bp

131-132 °C

density

0.809 g/mL at 20 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CC(C)CCO

InChI

1S/C5H12O/c1-5(2)3-4-6/h5-6H,3-4H2,1-2H3

InChI key

PHTQWCKDNZKARW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methylbutanol belongs to the group of high chain alcohols produced as metabolites from the degradation of amino acids via keto acids. It is a volatile flavor compound, highly present in alcoholic beverages.

Application

3-Methylbutanol may also be used as an analytical reference standard for the quantification of the analyte in liquor samples and milk samples using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

110.3 °F - closed cup

Flash Point(C)

43.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative determination of thermally derived off-flavor compounds in milk using solid-phase microextraction and gas chromatography
Vazquez-Landaverde A.P, et al.
Journal of Agricultural and Food Chemistry, 88(11), 3764-3772 (2005)
Flavours and Fragrances: Chemistry, Bioprocessing and Sustainability, 88(11), 3764-3772 (2007)
The
Wine Chemistry and Biochemistry, 88(11), 3764-3772 (2008)
Aroma perception of individual volatile compounds in fresh tomatoes (Lycopersicon esculentum, Mill.) as affected by the medium of evaluation
Tandon.S.K, et al.
Journal of Solution Chemistry, 20(3), 261-268 (2000)
Niels O Verhulst et al.
Malaria journal, 10, 28-28 (2011-02-10)
Anopheles gambiae sensu stricto is considered to be highly anthropophilic and volatiles of human origin provide essential cues during its host-seeking behaviour. A synthetic blend of three human-derived volatiles, ammonia, lactic acid and tetradecanoic acid, attracts A. gambiae. In addition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service