Merck
  • Electron-induced phase transition in hydrogen-bonded solid-state 2-pyridone.
Electron-induced phase transition in hydrogen-bonded solid-state 2-pyridone.
The journal of physical chemistry. A (2011-01-21)
Henryk T Flakus, Aleksandra Tyl, Andrzej Maślankiewicz
ABSTRACT
This paper presents the results of experimental studies of hydrogen-bonded 2-pyridone crystal IR spectra. Spectral studies have demonstrated the existence of two anhydrous solid-state phases of each compound, namely the α and the β phases. Hydrogen bonds in the high-temperature α phase of these crystals have been estimated to be 40% stronger than the hydrogen bonds in the β phase, which are stable at room temperature. The mechanism of the phase transition in the solid-state 2-pyridone is proposed on the basis of the IR spectral data. This was possible by taking into account small changes in the geometry of heterocyclic molecular skeletons, which accompany the electron density redistribution in the hydrogen bonds occurring during the transition. The phase transition is connected with a partial change in the hydrogen bond nature from the N(+)-H···O(-) in the α phase, to the N-H···O hydrogen bonds in the β phase crystals.
Sigma-Aldrich
2-Hydroxypyridine, 97%
Levetiracetam impurity C, European Pharmacopoeia (EP) Reference Standard