Skip to Content
Merck
All Photos(2)

Key Documents

H56800

Sigma-Aldrich

2-Hydroxypyridine

97%

Synonym(s):

2(1H)-Pyridone, 2-Pyridinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NO
CAS Number:
Molecular Weight:
95.10
Beilstein:
105757
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

280-281 °C (lit.)

mp

105-107 °C (lit.)

SMILES string

Oc1ccccn1

InChI

1S/C5H5NO/c7-5-3-1-2-4-6-5/h1-4H,(H,6,7)

InChI key

UBQKCCHYAOITMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for generating β-oxopropyl carbonates from cyclic carbonates and alcohols and in the aminolysis of a polyglutamate.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Journal of the Chemical Society. Perkin Transactions 1, 1749-1749 (1993)
Ali A Abdul-Sater et al.
Journal of immunology (Baltimore, Md. : 1950), 195(1), 210-216 (2015-05-29)
IFNs, which transduce pivotal signals through Stat1 and Stat2, effectively suppress the replication of Legionella pneumophila in primary murine macrophages. Although the ability of IFN-γ to impede L. pneumophila growth is fully dependent on Stat1, IFN-αβ unexpectedly suppresses L. pneumophila
Polymer, 35, 2443-2443 (1994)
Mettu Ravinder et al.
Bioorganic & medicinal chemistry letters, 22(18), 6010-6015 (2012-08-18)
Twenty-six 2-pyridone derivatives (8a-8z), which are structurally analogous to amrinone and milrinone two important cardiotonic drugs, are synthesized and characterized. The synthesis of 2-pyridone derivatives involves addition, followed by cyclization between Baylis-Hillman acetates (7a-7k) and enamino esters or nitriles (3a-3e).
Maria W Hirschbeck et al.
Structure (London, England : 1993), 20(1), 89-100 (2012-01-17)
The recently discovered FabV enoyl-ACP reductase, which catalyzes the last step of the bacterial fatty acid biosynthesis (FAS-II) pathway, is a promising but unexploited drug target against the reemerging pathogen Yersinia pestis. The structure of Y. pestis FabV in complex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service