Merck
Ionic liquid "buffers"-pH control in ionic liquid systems.
Chemical communications (Cambridge, England) (2010-09-28)
Douglas R Macfarlane, R Vijayaraghavan, Huy N Ha, Aleksey Izgorodin, Katherine D Weaver, Gloria D Elliott
ABSTRACT
We describe hydrated ionic liquids which have an intrinsic proton buffering action and hence an ability to control proton activity in an ionic medium.
Product Description
Supelco
Thymolphthalein, ACS reagent, indicator (pH 8.8-10.5)
Pricing and availability is not currently available.
Sigma-Aldrich
Thymol Blue, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Thymolphthalein, ACS reagent, Dye content 95 %
Pricing and availability is not currently available.
Sigma-Aldrich
Thymol Blue sodium salt, ACS reagent, Dye content 95 %
Pricing and availability is not currently available.