Skip to Content
Merck
All Photos(3)

Documents

114545

Sigma-Aldrich

Thymol Blue

ACS reagent

Synonym(s):

Thymolsulfonphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O5S
CAS Number:
Molecular Weight:
466.59
Beilstein:
368036
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

ACS reagent

Quality Level

form

powder

clarity of soln

passes test

technique(s)

titration: suitable

visual transition interval

1.2-2.8, red to yellow(acid range)
1.2-2.8, red to yellow
8.0-9.2, yellow to blue(alkaline range)
8.0-9.2, yellow to blue

mp

221-224 °C (dec.) (lit.)

solubility

ethanol: 0.1%, clear

density

0.979 g/cm3

λmax

376 nm (2nd)
594 nm

ε (extinction coefficient)

≥12000 at 298-302 nm in 0.1 M NaOH at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC(C)c1cc(c(C)cc1O)C2(OS(=O)(=O)c3ccccc23)c4cc(C(C)C)c(O)cc4C

InChI

1S/C27H30O5S/c1-15(2)19-13-22(17(5)11-24(19)28)27(21-9-7-8-10-26(21)33(30,31)32-27)23-14-20(16(3)4)25(29)12-18(23)6/h7-16,28-29H,1-6H3

InChI key

PRZSXZWFJHEZBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thymol Blue is a sulfonephthalein dye. It is also known as thymolsulfonphthalein.

Application

Thymol Blue has been used for the in situ spectrophotometric pH measurement.

Biochem/physiol Actions

Thymol Blue is a pH indicator, which is yellow/orange at low pH and blue in color at high pH.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Global stability for convection induced by absorption of radiation.
Straughan B
Dynamics of atmospheres and oceans, 35, 351-361 (2002)
N Takai et al.
Artificial organs, 15(2), 86-89 (1991-04-01)
An optical fiber sensor utilizing Thymol blue and an ion-exchange resin complex in a cellulose acetate membrane was developed. By monitoring several different chromophores of Thymol blue, the sensor could measure the pH of the solution from 1.0 to 12.0
S Passamonti et al.
Biochimica et biophysica acta, 943(2), 119-125 (1988-08-18)
Sulfobromophthalein electrogenic uptake into rat liver plasma membrane vesicles was shown to admit only the quinoid, trivalent anion. The minimum requirement for this electrogenic process has been investigated in rat liver plasma membrane vesicles by using Thymol blue, a pH-indicator
Elodie Verhaeghe et al.
Analytical biochemistry, 379(1), 60-65 (2008-05-22)
The standard assay for iodoperoxidase activity is based on the spectrophotometric detection of triiodide formed during the enzymatic reaction. However, chemical instability of I3- has limited the method to high iodide concentrations and acidic conditions. Here we describe a simple
Ali A Ensafi et al.
Journal of hazardous materials, 148(1-2), 319-325 (2007-03-30)
An on-line flow system was used to develop a selective and efficient on-line sorbent extraction preconcentration system for cadmium. The method is based on adsorption of cadmium ions onto the activated carbon modified with methyl thymol blue. Then the adsorbed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service