Merck
  • Peptide-catalyzed kinetic resolution of formamides and thioformamides as an entry to nonracemic amines.
Peptide-catalyzed kinetic resolution of formamides and thioformamides as an entry to nonracemic amines.
Journal of the American Chemical Society (2010-02-18)
Brandon S Fowler, Peter J Mikochik, Scott J Miller
ABSTRACT
We report a fundamentally unique approach to the catalytic kinetic resolution of amine derivatives based on formamide and thioformamide substrates. Readily accessible histidine-containing peptides mediate the kinetic resolutions with as little as 5 mol % catalyst. Selectivity factors (k(rel)) as high as 43.7 were observed under simple reaction conditions utilizing Boc(2)O as the reagent at room temperature. Mechanistic experiments were conducted that established a higher level of reactivity for thioformamide substrates than for their formamide analogues. The products of these asymmetric reactions were shown to be readily converted to desirable building blocks such as N-Boc-amines and the parent chiral formamide compounds.
Sigma-Aldrich
Di-tert-butyl dicarbonate, ReagentPlus®, ≥99%
Sigma-Aldrich
Di-tert-butyl dicarbonate, ≥98.0% (GC)
Sigma-Aldrich
Di-tert-butyl dicarbonate, ReagentPlus®, 99%
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 2 M in methylene chloride
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 2 M in THF
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 1.0 M in THF