Merck
  • Hypotensive and vasorelaxant effects of citronellol, a monoterpene alcohol, in rats.
Hypotensive and vasorelaxant effects of citronellol, a monoterpene alcohol, in rats.
Basic & clinical pharmacology & toxicology (2009-12-17)
Joana F A Bastos, Italo J A Moreira, Thaís P Ribeiro, Isac A Medeiros, Angelo R Antoniolli, Damião P De Sousa, Márcio R V Santos
ABSTRACT
Citronellol is an essential oil constituent from the medicinal plants Cymbopogon citratus, Cymbopogon winterianus and Lippia alba which are thought to possess antihypertensive properties. Citronellol-induced cardiovascular effects were evaluated in this study. In rats, citronellol (1-20 mg/kg, i.v.) induced hypotension, which was not affected by pre-treatment with atropine, hexamethonium, N(omega)-nitro-L-arginine methyl ester hydrochloride or indomethacin, and tachycardia, which was only attenuated by pre-treatment with atropine and hexamethonium. These responses were less than those obtained for nifedipine, a reference drug. In intact rings of rat mesenteric artery pre-contracted with 10 microM phenylephrine, citronellol induced relaxations (pD(2) = 0.71 +/- 0.11; E(max) = 102 +/- 5%; n = 6) that were not affected by endothelium removal, after tetraethylamonium in rings without endothelium pre-contracted with KCl 80 mM. Citronellol strongly antagonized (maximal inhibition = 97 +/- 4%; n = 6) the contractions induced by CaCl(2) (10(-6) to 3 x 10(-3 )M) and did not induce additional effects on the maximal response of nifedipine (10 microM). Finally, citronellol inhibited the contractions induced by 10 microM phenylephrine or 20 mM caffeine. The present results suggest that citronellol lowers blood pressure by a direct effect on the vascular smooth muscle leading to vasodilation.
Sigma-Aldrich
(R)-(+)-β-Citronellol, 95%
Supelco
(−)-β-Citronellol, analytical standard
Sigma-Aldrich
(S)-(−)-β-Citronellol, ≥97%
Sigma-Aldrich
Citronellol, ≥95%, FCC, FG
Sigma-Aldrich
β-Citronellol, 95%
Supelco
(±)-β-Citronellol, analytical standard
Sigma-Aldrich
Citronellol, mixture of isomers, natural, ≥95%, FG