Skip to Content
Merck
All Photos(1)

Key Documents

303488

Sigma-Aldrich

(S)-(−)-β-Citronellol

≥97%

Synonym(s):

β-Rhodinol, (S)-3,7-Dimethyl-6-octen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
156.27
Beilstein:
1721505
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

~0.02 mmHg ( 25 °C)

Quality Level

Assay

≥97%

form

liquid

optical activity

[α]20/D −4.5°, neat

refractive index

n20/D 1.456 (lit.)

bp

225-226 °C (lit.)

density

0.856 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

C[C@H](CCO)CC\C=C(\C)C

InChI

1S/C10H20O/c1-9(2)5-4-6-10(3)7-8-11/h5,10-11H,4,6-8H2,1-3H3/t10-/m0/s1

InChI key

QMVPMAAFGQKVCJ-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renan G Brito et al.
Basic & clinical pharmacology & toxicology, 112(4), 215-221 (2012-10-06)
Citronellol (CT) is a monoterpenoid alcohol present in the essential oil of many medicinal plants, such as Cymbopogon citratus. We evaluated the antinociceptive effects of CT on orofacial nociception in mice and investigated the central pathway involved in the effect.
Ram Swaroop Verma et al.
Natural product research, 25(17), 1577-1584 (2011-06-30)
Rosa damascena Mill. is an important aromatic plant for commercial production of rose oil, water, concrete and absolute. The rose water and rose oil produced under the mountainous conditions of Uttarakhand were investigated for their chemical composition. The major components
Shalinder Kaur et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 66(5-6), 260-266 (2011-08-05)
Citronellol, an oxygenated monoterpene, is found naturally in the essential oils of several aromatic plants and has been reported to exhibit growth inhibitory and pesticidal activities. However, its mechanism of action is largely unexplored. We investigated the effect of citronellol
Wantida Chaiyana et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(8-9), 836-839 (2012-04-19)
Inhibition of cholinesterase has attracted much attention recently because of its potential for the treatment of Alzheimer's disease. In this work, the anticholinesterase activities of plant oils were investigated using Ellman's colorimetric method. The results indicate that essential oils obtained
Yoshikazu Kitano et al.
Biofouling, 27(2), 201-205 (2011-02-01)
Twenty novel simple alkyl isocyanides derived from citronellol were synthesized and evaluated for their antifouling activity and toxicity against cypris larvae of the barnacle, Balanus amphitrite. The anti-barnacle activity of the synthesized isocyanides was in the EC(50) range of 0.08-1.49

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service