Merck
  • End-functionalised 1-vinyl-2-pyrrolidinone oligomers bearing lactate functions at one end.
End-functionalised 1-vinyl-2-pyrrolidinone oligomers bearing lactate functions at one end.
Macromolecular bioscience (2004-10-07)
Elisabetta Ranucci, Luca Macchi, Rita Annunziata, Paolo Ferruti, Federica Chiellini
ABSTRACT
1-Vinyl-2-pyrrolidinone (VP) oligomers bearing a lactate group at one end (PVP-L) were obtained by chain-transfer controlled radical polymerisation carried out in the presence of ethyl L-lactate as chain-transfer agent (CTA). Their number-average molecular weights were in the range 1500-4000 with molecular weight distributions ranging from 1.4 to 1.8. The chain transfer constant, C(T), of the ethyl L-lactate/VP system was determined by monitoring the variation of PVP-L number-average molecular weight on conversion. The C(T) value so obtained was 1.03 x 10(-2), which is by about one order of magnitude higher than the C(T) value previously determined for a seemingly similar system, namely methyl isobutyrate/1-vinyl-2-pyrrolidinone (1.64 x 10(-3)). The resultant PVP-L oligomers were thoroughly characterised by means of (1)H and (13)C NMR, in order to ascertain the regular presence of the lactate functions at one of their chain terminals. NMR characterisations gave results in full agreement with the proposed structure. Moreover, the molecular weight values determined by NMR very closely agreed with those obtained by SEC. Preliminary biological evaluations of the PVP-L oligomers showed a complete lack of toxicity.
Sigma-Aldrich
(−)-Ethyl L-lactate, ≥99.0%
Sigma-Aldrich
Ethyl lactate, ≥98%, FCC, FG
Sigma-Aldrich
(−)-Ethyl L-lactate, purum, ≥98.0% (sum of enantiomers, GC)
Sigma-Aldrich
Ethyl lactate, natural, ≥98%, FCC, FG
Sigma-Aldrich
(−)-Ethyl L-lactate, 98%