Skip to Content
Merck
All Photos(2)

Key Documents

E34102

Sigma-Aldrich

(−)-Ethyl L-lactate

98%

Synonym(s):

(−)-Ethyl (S)-2-hydroxypropionate, (S)-(−)-2-Hydroxypropionic acid ethyl ester, L-Lactic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O3
CAS Number:
Molecular Weight:
118.13
Beilstein:
1720839
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.1 (vs air)

Quality Level

vapor pressure

2 mmHg ( 20 °C)
5 mmHg ( 30 °C)

Assay

98%

form

liquid

optical activity

[α]14/D −10°, neat

autoignition temp.

752 °F

expl. lim.

1.5 %, 100 °F

refractive index

n20/D 1.4130 (lit.)

pH

4 (20 °C, 50 g/L)

bp

154 °C (lit.)

mp

−26 °C (lit.)

density

1.034 g/mL at 20 °C (lit.)
1.042 g/mL at 25 °C

SMILES string

C[C@H](O)C(OCC)=O

InChI

1S/C5H10O3/c1-3-8-5(7)4(2)6/h4,6H,3H2,1-2H3/t4-/m0/s1

InChI key

LZCLXQDLBQLTDK-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl lactate is an economically viable green solvent used in organic synthesis. It is generally employed in organic reactions like multi-component reactions, cycloaddition reaction, asymmetric induction, photochemical synthesis, and ligand-free coupling reactions, etc.

Application

(−)-Ethyl L-lactate can be used in the preparation of:
  • (R)-(+)-2-methyloxirane.
  • (S)-2-((tert-Butyldimethylsilyl)oxy)propanal, a key intermediate for the synthesis of maltepolides.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

128.8 °F - closed cup

Flash Point(C)

53.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethyl lactate as a tunable solvent for the synthesis of aryl aldimines
Bennett J, et al.
Green Chemistry, 11(2), 166-168 (2009)
Ethyl lactate as a green solvent: a promising bio-compatible media for organic synthesis
Paul S, et al.
Current Green Chemistry, 3(1), 111-118 (2016)
A facile approach for the synthesis of C13-C24 fragments of maltepolides A, C and D
Rao, P Sankara and Srihari, P
Organic & Biomolecular Chemistry, 14(40), 9629-9638 (2016)
Shuke Xiao et al.
eLife, 8 (2019-10-28)
Odorant binding proteins (Obps) are expressed at extremely high levels in the antennae of insects, and have long been believed essential for carrying hydrophobic odorants to odor receptors. Previously we found that when one functional type of olfactory sensillum in
Eduardo Coelho et al.
Molecules (Basel, Switzerland), 25(3) (2020-02-07)
Knowledge of composition of beverages volatile fraction is essential for understanding their sensory attributes. Analysis of volatile compounds predominantly resorts to gas chromatography coupled with mass spectrometry (GC-MS). Often a previous concentration step is required to quantify compounds found at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service