Merck
  • Flow cytometric in situ proximity ligation analyses of protein interactions and post-translational modification of the epidermal growth factor receptor family.
Flow cytometric in situ proximity ligation analyses of protein interactions and post-translational modification of the epidermal growth factor receptor family.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2009-08-04)
Karl-Johan Leuchowius, Irene Weibrecht, Ulf Landegren, Lars Gedda, Ola Söderberg
ABSTRACT
Interactions between members of the epidermal growth factor receptor (EGFR) family mediates cellular responses to ligand stimulation. Measurement of these interactions could provide important information and may prove useful as prognostic markers in malignancy. Therefore, to develop methods to study these interactions in genetically unmodified cells, such as clinical samples, in a sensitive and selective way, with good statistical accuracy, is important. The in situ proximity ligation assay (in situ PLA) was used to quantify homo- and heteromeric interactions between EGFR and HER2 in cultured cells, using flow cytometry as the readout method. Cells were monitored for changes in dimerization patterns and phosphorylation status upon stimulation. The different cell lines displayed varying amounts of interactions between EGFR and HER2, but the amount of dimerization was not found to be affected significantly upon stimulation by EGF. Activation of EGFR could be visualized by in situ PLA, but not by immunofluorescence staining. In situ PLA was successfully used to study receptor dimerization and activation of the EGF-receptor family with high selectivity and sensitivity. The combination of in situ PLA and flow cytometry provided a statistically powerful way of analyzing protein-protein interactions and post-translational modifications on a single-cell basis.
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection