Merck
  • IL11 antagonist inhibits uterine stromal differentiation, causing pregnancy failure in mice.
IL11 antagonist inhibits uterine stromal differentiation, causing pregnancy failure in mice.
Biology of reproduction (2009-01-16)
Ellen Menkhorst, Lois Salamonsen, Lorraine Robb, Evdokia Dimitriadis
ABSTRACT
Hormonal contraceptives are unsuitable for many women; thus, the development of new, nonhormonal contraceptives is of great interest. In women, uterine epithelial expression of interleukin 11 (IL11) and its receptor (IL11RA) suggests IL11 is critical for blastocyst attachment during implantation. Il11ra-deficient mice are infertile due to a defective decidualization response to the blastocyst, leading to total pregnancy loss. We examined the effect of administering a PEGylated IL11 antagonist, PEGIL11A (where PEG is polyethylene glycol), on pregnancy outcomes in mice and IL11 signaling in human endometrial epithelial cells (HES). PEGIL11A was detected in sera up to 72 h after intraperitoneal (IP) injection versus up to 2 h for the non-PEGylated antagonist. Following IP injection, PEGIL11A localized to uterine decidual cells and reduced immunoreactive cyclin D3 (IL11 decidual target). To inhibit IL11 action during early decidualization, PEGIL11A or control were administered IP on Days 3-6 (beginning just prior to maximal decidual Il11 expression). On Day 6, mesometrial decidualization was disturbed in PEGIL11A-treated animals with regions of hemorrhage visible in the mesometrial decidua. On Day 10, severe decidual destruction was visible: implantation sites contained significant hemorrhage, and the uterine luminal epithelium had reformed, suggesting a return to estrous cycling. These results demonstrate that PEGIL11A blocked IL11 action in the decidua during early decidualization, which totally abolished pregnancy and which is equivalent to the Il11ra(-/-) mouse. PEGIL11A significantly diminished STAT3 phosphorylation in HES cells in vitro (P < or = 0.05). This study provides valuable information for PEGIL11A that could lead to the development of this protein as a nonhormonal contraceptive.
Sigma-Aldrich
Methyl Blue, certified by the BSC
Sigma-Aldrich
Methyl Blue, powder or crystals
Sigma-Aldrich
Methyl Blue, for microscopy (Bot., Hist.), indicator (pH 9.4-14.0)
Sigma-Aldrich
StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, AutoMod, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
SAFC
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications