Merck
H3K4 methylation by SETD1A/BOD1L facilitates RIF1-dependent NHEJ.
Molecular cell (2022-04-20)
Rachel Bayley, Valerie Borel, Rhiannon J Moss, Ellie Sweatman, Philip Ruis, Alice Ormrod, Amalia Goula, Rachel M A Mottram, Tyler Stanage, Graeme Hewitt, Marco Saponaro, Grant S Stewart, Simon J Boulton, Martin R Higgs
ABSTRACT
The 53BP1-RIF1-shieldin pathway maintains genome stability by suppressing nucleolytic degradation of DNA ends at double-strand breaks (DSBs). Although RIF1 interacts with damaged chromatin via phospho-53BP1 and facilitates recruitment of the shieldin complex to DSBs, it is unclear whether other regulatory cues contribute to this response. Here, we implicate methylation of histone H3 at lysine 4 by SETD1A-BOD1L in the recruitment of RIF1 to DSBs. Compromising SETD1A or BOD1L expression or deregulating H3K4 methylation allows uncontrolled resection of DNA ends, impairs end-joining of dysfunctional telomeres, and abrogates class switch recombination. Moreover, defects in RIF1 localization to DSBs are evident in patient cells bearing loss-of-function mutations in SETD1A. Loss of SETD1A-dependent RIF1 recruitment in BRCA1-deficient cells restores homologous recombination and leads to resistance to poly(ADP-ribose)polymerase inhibition, reinforcing the clinical relevance of these observations. Mechanistically, RIF1 binds directly to methylated H3K4, facilitating its recruitment to, or stabilization at, DSBs.
Sigma-Aldrich
Anti-Chk1 antibody, Mouse monoclonal, ~2 mg/mL, clone DCS-310, purified from hybridoma cell culture
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-Histone H2A (acidic patch) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-PTIP Antibody, from chicken, purified by affinity chromatography
Sigma-Aldrich
Anti-Rad51 (Ab-1) Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Anti-Chk2 Antibody, clone 7, clone 7, Upstate®, from mouse
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys4) Antibody, clone AW30, rabbit monoclonal, culture supernatant, clone AW30, Upstate®
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Sigma-Aldrich
Hydroxyurea, 98%, powder