Merck
  • SH3BP4 promotes neuropilin-1 and α5-integrin endocytosis and is inhibited by Akt.
SH3BP4 promotes neuropilin-1 and α5-integrin endocytosis and is inhibited by Akt.
Developmental cell (2021-03-25)
Christoph J Burckhardt, John D Minna, Gaudenz Danuser
ABSTRACT
Cells probe their surrounding matrix for attachment sites via integrins that are internalized by endocytosis. We find that SH3BP4 regulates integrin surface expression in a signaling-dependent manner via clathrin-coated pits (CCPs). Dephosphorylated SH3BP4 at S246 is efficiently recruited to CCPs, while upon Akt phosphorylation, SH3BP4 is sequestered by 14-3-3 adaptors and excluded from CCPs. In the absence of Akt activity, SH3BP4 binds GIPC1 and targets neuropilin-1 and α5/β1-integrin for endocytosis, leading to inhibition of cell spreading. Similarly, chemorepellent semaphorin-3a binds neuropilin-1 to activate PTEN, which antagonizes Akt and thus recruits SH3BP4 to CCPs to internalize both receptors and induce cell contraction. In PTEN mutant non-small cell lung cancer cells with high Akt activity, expression of non-phosphorylatable active SH3BP4-S246A restores semaphorin-3a induced cell contraction. Thus, SH3BP4 links Akt signaling to endocytosis of NRP1 and α5/β1-integrins to modulate cell-matrix interactions in response to intrinsic and extrinsic cues.
Product Description
Roche
Streptavidin-POD conjugate, liquid, pkg of 1 mL
Sigma-Aldrich
Anti-HA-Tag antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
MISSION® esiRNA, targeting human CLTC
Sigma-Aldrich
Anti-Integrin alpha5 (Preservative Free) Antibody, clone SNAKA51, clone SNAKA51, from mouse, purified by using protein G
Sigma-Aldrich
Blood-Brain Barrier hCMEC/D3 Cell Line, The hCMEC/D3 BBB cell line has been extensively characterized for brain endothelial phenotype and is a model of human blood-brain barrier (BBB) function.
Sigma-Aldrich
Anti-Integrin alpha 5 (CD49e) Antibody, clone mAb11, clone mAB11, from rat
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Guanidine thiocyanate, for molecular biology, ≥99%
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone V9, ascites fluid
Sigma-Aldrich
DAPI, for nucleic acid staining
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)