Merck
  • Measuring CPT-1-mediated respiration in permeabilized cells and isolated mitochondria.
Measuring CPT-1-mediated respiration in permeabilized cells and isolated mitochondria.
STAR protocols (2021-08-18)
Krista Yang, Mary T Doan, Linsey Stiles, Ajit S Divakaruni
ABSTRACT
Carnitine palmitoyltransferase-1 (CPT-1) is a rate-controlling enzyme for long-chain fatty acid oxidation. This manuscript provides protocols for measuring CPT-1-mediated respiration in permeabilized, adherent cell monolayers and mitochondria freshly isolated from tissue, along with examples to assess the potency and specificity of interventions targeting CPT-1. Strengths of the approach include ease, speed, and breadth of analysis, whereas drawbacks include loss of physiological regulation in reductionist systems and indirect assessment of CPT-1 enzymatic activity. For complete details on the use and execution of this protocol, please refer to Divakaruni et al. (2018).
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Coenzyme A sodium salt hydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Palmitoyl coenzyme A lithium salt, ≥90%
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Adenosine 5′-diphosphate monopotassium salt dihydrate, bacterial, ≥95%, powder
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Sigma-Aldrich
Pyruvic acid, 98%
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Succinic acid, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
sn-Glycerol 3-phosphate bis(cyclohexylammonium) salt, ≥93% (GC)
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Palmitoyl-L-carnitine, ≥97.0% (HPLC)
Sigma-Aldrich
Calcium chloride solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Sucrose, BioUltra, ≥99.5% (HPLC)