Skip to Content
Merck
All Photos(1)

Key Documents

61251

Sigma-Aldrich

Palmitoyl-L-carnitine

≥97.0% (HPLC)

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxohexadecyl)oxy]-1-propanaminium inner salt, L-Carnitine hexadecanoyl ester, C16-Carnitine, Hexadecanoyl-L-carnitine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H45NO4
CAS Number:
Molecular Weight:
399.61
Beilstein:
4152034
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥97.0% (HPLC)

optical activity

[α]/D -17±2°, c = 1 in methanol

impurities

≤10% water

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](OC(CCCCCCCCCCCCCCC)=O)CC([O-])=O

InChI

1S/C23H45NO4/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(27)28-21(19-22(25)26)20-24(2,3)4/h21H,5-20H2,1-4H3/t21-/m1/s1

InChI key

XOMRRQXKHMYMOC-OAQYLSRUSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-Palmitoylcarnitine can change the activity of several enzymes and transporters, localized in the membrane and facilitates the transfer of long-chain fatty acids from cytoplasm into mitochondria during the oxidation of fatty acids. L-Palmitoylcarnitine accumulates in ischemic myocardium and potentially contribute to myocardial damage through alterations in membrane molecular dynamics.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katherine A Overmyer et al.
Cell metabolism, 21(3), 468-478 (2015-03-05)
Maximal exercise-associated oxidative capacity is strongly correlated with health and longevity in humans. Rats selectively bred for high running capacity (HCR) have improved metabolic health and are longer-lived than their low-capacity counterparts (LCR). Using metabolomic and proteomic profiling, we show
Sanne J C M Frambach et al.
Biochimica et biophysica acta. Molecular basis of disease, 1866(6), 165727-165727 (2020-02-20)
Mitochondrial complex I (CI), the first multiprotein enzyme complex of the OXPHOS system, executes a major role in cellular ATP generation. Consequently, dysfunction of this complex has been linked to inherited metabolic disorders, including Leigh disease (LD), an often fatal
Ji Young Kim et al.
Journal of proteome research, 9(9), 4368-4375 (2010-06-22)
Obesity is currently epidemic in many countries worldwide and is strongly related to diabetes and cardiovascular disease. This study investigated the differences in metabolomic profiling between overweight/obese and normal-weight men. Overweight/obese (n=30) and age-matched, normal-weight men (n=30) were included. Anthropometric
Adolfas Toleikis et al.
Cells, 9(2) (2020-02-07)
Fatty acids are the main respiratory substrates important for cardiac function, and their oxidation is altered during various chronic disorders. We investigated the mechanism of fatty acid-oxidation-induced changes and their relations with mitochondrial morphology and ADP/ATP carrier conformation on the
Stephanie J Mihalik et al.
Obesity (Silver Spring, Md.), 18(9), 1695-1700 (2010-01-30)
Dysregulation of fatty acid oxidation (FAO) is recognized as important in the pathophysiology of obesity and insulin resistance (IR). However, demonstrating FAO defects in vivo in humans has entailed complex and invasive methodologies. Recently, the identification of genetic blocks in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service