Merck
  • KAI1(CD82) is a key molecule to control angiogenesis and switch angiogenic milieu to quiescent state.
KAI1(CD82) is a key molecule to control angiogenesis and switch angiogenic milieu to quiescent state.
Journal of hematology & oncology (2021-09-18)
Jin-Woo Lee, Jin Hur, Yoo-Wook Kwon, Cheong-Whan Chae, Jae-Il Choi, Injoo Hwang, Ji-Yeon Yun, Jin-A Kang, Young-Eun Choi, Young Hyun Kim, Sang Eun Lee, Cheol Lee, Dong Hyun Jo, Heeyoung Seok, Byong Seung Cho, Sung Hee Baek, Hyo-Soo Kim
ABSTRACT
Little is known about endogenous inhibitors of angiogenic growth factors. In this study, we identified a novel endogenous anti-angiogenic factor expressed in pericytes and clarified its underlying mechanism and clinical significance. Herein, we found Kai1 knockout mice showed significantly enhanced angiogenesis. Then, we investigated the anti-angiogenic roll of Kai1 in vitro and in vivo. KAI1 was mainly expressed in pericytes rather than in endothelial cells. It localized at the membrane surface after palmitoylation by zDHHC4 enzyme and induced LIF through the Src/p53 pathway. LIF released from pericytes in turn suppressed angiogenic factors in endothelial cells as well as in pericytes themselves, leading to inhibition of angiogenesis. Interestingly, KAI1 had another mechanism to inhibit angiogenesis: It directly bound to VEGF and PDGF and inhibited activation of their receptors. In the two different in vivo cancer models, KAI1 supplementation significantly inhibited tumor angiogenesis and growth. A peptide derived from the large extracellular loop of KAI1 has been shown to have anti-angiogenic effects to block the progression of breast cancer and retinal neovascularization in vivo. KAI1 from PC is a novel molecular regulator that counterbalances the effect of angiogenic factors.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia), FITC conjugate, lyophilized powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-PDGF-BB Antibody, from rabbit, purified by affinity chromatography
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Collagen Type IV Antibody, Chemicon®, from rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-NG2 Chondroitin Sulfate Proteoglycan, Cy3 Conjugate Antibody, from rabbit, CY3 conjugate
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, clone 4A6, Upstate®, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-VEGF Antibody, from rabbit, purified by affinity chromatography
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-p53 (pantropic) Antibody, clone DO-1, clone DO-1, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
PP2, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-NG2, Alexa Fluor488 Conjugate Antibody, from rabbit, ALEXA FLUOR 488