Merck
  • Suppressing Subcapsular Sinus Macrophages Enhances Transport of Nanovaccines to Lymph Node Follicles for Robust Humoral Immunity.
Suppressing Subcapsular Sinus Macrophages Enhances Transport of Nanovaccines to Lymph Node Follicles for Robust Humoral Immunity.
ACS nano (2020-06-02)
Yi-Nan Zhang, Wilson Poon, Elana Sefton, Warren C W Chan
ABSTRACT
Nanovaccines need to be transported to lymph node follicles to induce humoral immunity and generate neutralizing antibodies. Here, we discovered that subcapsular sinus macrophages play a barrier role to prevent nanovaccines from accessing lymph node follicles. This is illustrated by measuring the humoral immune responses after removing or functionally altering these cells in the nanovaccine transport process. We achieved up to 60 times more antigen-specific antibody production after suppressing subcapsular sinus macrophages. The degree of the enhanced antibody production is dependent on the nanovaccine dose and size, formulation, and administration time. We further found that pharmacological agents that disrupt the macrophage uptake function can be considered as adjuvants in vaccine development. Immunizing mice using nanovaccines formulated with these agents can induce more than 30 times higher antigen-specific antibody production compared to nanovaccines alone. These findings suggest that altering transport barriers to enable more of the nanovaccine to be delivered to the lymph node follicles for neutralizing antibody production is an effective strategy to boost vaccination.
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
ι-Carrageenan, commercial grade, Predominantly iota carrageenan
Sigma-Aldrich
Casein Blocking Buffer 10x, for Northern and Southern blotting, solution
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Gadolinium(III) chloride hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., average mol wt >500,000 (dextran starting material), contains 0.5-2.0% phosphate buffer, pH 6-8
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., average mol wt 9,000-20,000
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Adiponitrile, battery grade, >99%, acid <200 ppm, H2O <100 ppm