Merck
Synthesis and antioxidant properties of dendritic polyphenols.
Bioorganic & medicinal chemistry letters (2009-10-14)
Choon Young Lee, Ajit Sharma, Jae Eun Cheong, Julie L Nelson
ABSTRACT
Three dendritic polyphenols (generation 1) were synthesized: a syringaldehyde-based dendrimer (1), a vanillin-based dendrimer (2), and an iodinated vanillin-based dendrimer (3). They all showed strong antioxidant activity according to the 1,1-diphenyl-2-picrylhydrazyl (DPPH) radical assay. The syringaldehyde dendrimer was twice and 10 times stronger than quercetin and Trolox, respectively. The vanillin-based dendrimer and its more hydrophobic iodinated derivative were also more potent antioxidants than quercetin and Trolox. The DPPH order of potency was 1>2, 3>quercetin>Trolox. All three dendrimers also protected human LDL from free radical attack in a dose-dependent manner. Their order of free radical scavenging was 1>3>2>quercetin>Trolox. The increased hydrophobic nature of the iodinated derivative may have contributed to its better LDL protection than 2. Protection of linoleic acid oxidation was studied by the beta-carotene-linoleate assay. Dendrimer 1 was clearly superior to the other antioxidants in protecting the fatty acid. In case of DNA protection against free radical damage, the order of activity was 1>quercetin>2>3, Trolox. Pro-oxidant effect on copper-induced DNA oxidation showed the following order: quercetin, Trolox>1>2>3. Results of the study show that dendritic antioxidants, even at the generation 1 level, provide promising antioxidant properties for their potential use as drug candidates for diseases associated with oxidative stress.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
p-Xylylenediamine, 99%
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Syringaldehyde, ≥98%, FG