Merck
  • Quantitative structure-activity relationship analysis of inhibitors of the nicotine metabolizing CYP2A6 enzyme.
Quantitative structure-activity relationship analysis of inhibitors of the nicotine metabolizing CYP2A6 enzyme.
Journal of medicinal chemistry (2005-01-22)
Minna Rahnasto, Hannu Raunio, Antti Poso, Carsten Wittekindt, Risto O Juvonen
ABSTRACT
The purpose of this study was to develop screening and in silico modeling methods to obtain accurate information on the active center of CYP2A6, a nicotine oxidizing enzyme. The inhibitory potencies of 26 naphthalene and 16 non-naphthalene derivatives were determined for human CYP2A6 and mouse CYP2A5 enzymes. Several comparative molecular field analysis (CoMFA) models were developed to find out what types of steric and electrostatic properties are required for potent inhibitors. The IC(50) values of the tested compounds varied from 0.55 to 35 400 microM for CYP2A6 and from 1 to 1500 microM for CYP2A5. The generated CoMFA models were able to accurately predict the inhibition potencies of an external test set of chemicals. Potent and specific inhibitors of the CYP2A6 enzyme can be used in the future to increase nicotine bioavailability and thus make oral nicotine administration feasible in smoking cessation therapy.
Sigma-Aldrich
1-Methylnaphthalene, 95%
Supelco
Naphthalene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
p-Tolualdehyde, ≥97%, FG
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
3-Methylisoquinoline, 98%
Sigma-Aldrich
1,3-Dimethylnaphthalene, 96%
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
1,6-Dimethylnaphthalene, 99%
Sigma-Aldrich
α-Tetralone, 97%
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
1,2-Dimethylnaphthalene, 95%
Sigma-Aldrich
Phenethylamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
1-Methylnaphthalene, ≥95%
Sigma-Aldrich
1-Chloronaphthalene, technical grade
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
2,6-Dimethylnaphthalene, 99%
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Sigma-Aldrich
(−)-Cotinine, ≥98%
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Supelco
(−)-Cotinine solution, drug standard, 1.0 mg/mL in methanol
Sigma-Aldrich
Phenethylamine, ≥99%
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Supelco
1-Methylnaphthalene, analytical standard
Supelco
Naphthalene, analytical standard
Sigma-Aldrich
1-Chloronaphthalene, technical, ≥85% (GC)