Merck
Toxicological analysis of low-nicotine and nicotine-free cigarettes.
Toxicology (2008-07-05)
Jinguo Chen, Richard Higby, Defa Tian, Duanjun Tan, Michael D Johnson, Yingxian Xiao, Kenneth J Kellar, Shibao Feng, Peter G Shields
ABSTRACT
Low-nicotine and nicotine-free cigarettes are commercially available under the brand-name Quest. Some consumers may believe that these are safer cigarettes, and they may smoke more cigarettes or inhale more smoke to compensate for low nicotine yields. Thus, we have studied the toxicological effects of these two cigarettes and compared them with the Kentucky reference cigarette 2R4F. Also, the availability of nicotine-free cigarettes allows for the assessing the role of nicotine in cigarette smoke. In addition to nicotine, some tobacco-specific nitrosamines, aldehydes, and volatile organic compounds were also reduced in the Quest cigarettes compared to the 2R4F. However, aromatic amines were higher in the nicotine-free compared with low nicotine cigarettes. The Ames test revealed that cigarette smoke condensates from the nicotine-free (CSC-F), low nicotine (CSC-L) and 2R4F (CSC-R) cigarettes had a similar mutagenic potency. Exposure to any CSC caused a similar dose-dependent LDH leakage from normal human bronchial epithelial cells. However, CSC-F had more inhibitory effects on the cell growth than CSC-L and CSC-R. Adding nicotine to the CSC-F attenuated this inhibition. Both Quest CSCs decreased gap junction intercellular communication and caused cell cycle arrest. CSC exposure increased cytoplasmic nucleosomes, sub-G1/G0 population and apoptotic comet tails. Proapoptotic protein Bax increased independent of p53 induction after exposure to CSC-F. In conclusion, these studies are not consistent with a perception that low-nicotine or nicotine-free cigarettes may have less toxicity in human cells. Nicotine, as it exists in CSC, attenuates cytotoxicity possibly in part through inhibition of apoptotic pathways.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
1-Heptanol, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
1-Heptanol, 98%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
PKH26 Reference Microbeads
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
1-Heptanol, analytical standard, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%