Merck
Optimizing multiplex and LA-PCR with betaine.
Trends in biochemical sciences (1997-06-01)
P N Hengen
ABSTRACT
Methods and reagents is a unique monthly column that highlights current discussions in the newsgroup bionet.molbio.methds-reagnts, available on the internet. This month's column discusses the use of additives for optimizing the amount and quality of product obtained through multiplex and 'long and accurate' polymerase chain reaction (LA-PCR). For details on how to partake in the newsgroup, see the accompanying box.
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent