Merck
  • Strategy used to synthesize high activity and low Pd catalyst for Suzuki coupling reaction: an experimental and theoretical investigation.
Strategy used to synthesize high activity and low Pd catalyst for Suzuki coupling reaction: an experimental and theoretical investigation.
Physical chemistry chemical physics : PCCP (2020-03-05)
Xiao Yan, Yafei Luo, Wei Liu, Li Liang, Ya Gan, Zhongzhu Chen, Zhigang Xu, Hua Wan, Dianyong Tang, Hubing Shi, Jianping Hu
ABSTRACT
Unveiling the reaction mechanism is significant for developing high-performance catalysts. In this paper, a series of precisely controlled PdxM147-x (M = Cu, Pt, Au, Rh, Ru) dendrimer encapsulated nanoparticles (DENs) has been successfully synthesized. The mechanisms of PdxM147-x as catalysts for Suzuki cross-coupling reactions were investigated by combining experimental and theoretical methods. The experimental results indicate that Pd74Cu73 DEN shows similar activity to Pd147 DEN and excellent substrate adaptability under mild reaction conditions. Moreover, the Cu component can play an important role in tuning the catalytic activity of PdxCu147-x DEN. Density functional theory (DFT) calculations illustrate that the similar activities of the Pd147 and Pd74Cu73 DENs originate from the comparable energy barriers of the rate-determining steps. The partial density of states (PDOS) and electron density differences demonstrate that Cu decreases the intensities of the valence orbitals of the top and edge Pd atoms and weakens orbital interactions between the intermediates and Pd74Cu73 DEN, leading to low desorption energies of the products. This work can provide a promising strategy to reduce the cost of Pd catalysts in Suzuki cross-coupling reactions.
Sigma-Aldrich
4-Bromobenzotrifluoride, 99%
Sigma-Aldrich
1-Bromo-4-nitrobenzene, 99%
Sigma-Aldrich
4-(Trifluoromethyl)phenylboronic acid, ≥95.0%
Sigma-Aldrich
4-Bromoanisole, ≥99.0%
Sigma-Aldrich
4-Methoxyphenylboronic acid, ≥95.0%
Sigma-Aldrich
1-Bromo-4-tert-butylbenzene, 97%
Sigma-Aldrich
4-tert-Butylphenylboronic acid, ≥95.0%
Sigma-Aldrich
4-Chlorophenylboronic acid, 95%