Merck
ATP13A2 deficiency disrupts lysosomal polyamine export.
Nature (2020-01-31)
Sarah van Veen, Shaun Martin, Chris Van den Haute, Veronick Benoy, Joseph Lyons, Roeland Vanhoutte, Jan Pascal Kahler, Jean-Paul Decuypere, Géraldine Gelders, Eric Lambie, Jeffrey Zielich, Johannes V Swinnen, Wim Annaert, Patrizia Agostinis, Bart Ghesquière, Steven Verhelst, Veerle Baekelandt, Jan Eggermont, Peter Vangheluwe
ABSTRACT
ATP13A2 (PARK9) is a late endolysosomal transporter that is genetically implicated in a spectrum of neurodegenerative disorders, including Kufor-Rakeb syndrome-a parkinsonism with dementia1-and early-onset Parkinson's disease2. ATP13A2 offers protection against genetic and environmental risk factors of Parkinson's disease, whereas loss of ATP13A2 compromises lysosomes3. However, the transport function of ATP13A2 in lysosomes remains unclear. Here we establish ATP13A2 as a lysosomal polyamine exporter that shows the highest affinity for spermine among the polyamines examined. Polyamines stimulate the activity of purified ATP13A2, whereas ATP13A2 mutants that are implicated in disease are functionally impaired to a degree that correlates with the disease phenotype. ATP13A2 promotes the cellular uptake of polyamines by endocytosis and transports them into the cytosol, highlighting a role for endolysosomes in the uptake of polyamines into cells. At high concentrations polyamines induce cell toxicity, which is exacerbated by ATP13A2 loss due to lysosomal dysfunction, lysosomal rupture and cathepsin B activation. This phenotype is recapitulated in neurons and nematodes with impaired expression of ATP13A2 or its orthologues. We present defective lysosomal polyamine export as a mechanism for lysosome-dependent cell death that may be implicated in neurodegeneration, and shed light on the molecular identity of the mammalian polyamine transport system.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-ATP13A2 (C-terminal region) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Resomer® RG 503 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 24,000-38,000
Sigma-Aldrich
Dynasore hydrate
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Histamine dihydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Spermidine trihydrochloride, ≥98% (TLC)
Sigma-Aldrich
4-Methylumbelliferyl heptanoate, ≥95% (GC)
Sigma-Aldrich
Agmatine sulfate salt, ≥97%, powder
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Yeast Synthetic Drop-out Medium Supplements, without uracil
Supelco
Bradford Reagent, for 0.1-1.4 mg/ml protein
Sigma-Aldrich
Spermine, ≥97%
Sigma-Aldrich
BioTracker Spermidine Polyamine Green Fluorescent Live Cell Probe
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
BioTracker Putrescine Polyamine Green Fluorescent Live Cell Probe
Streptavidin Sepharose High Performance, Cytiva 17-5113-01, pack of 5 mL
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Cadaverine, 95%
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 70,000, (FITC:Glucose = 1:250)
Sigma-Aldrich
L-Ornithine monohydrochloride, ≥99%
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
CA-074, ≥98% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Putrescine dihydrochloride, ≥98% (TLC)
Supelco
N8-Acetylspermidine dihydrochloride, analytical standard
Thrombin Protease, Cytiva 27-0846-01
Sigma-Aldrich
Pitstop 2, ≥98% (HPLC)