Merck
Resilient living materials built by printing bacterial spores.
Nature chemical biology (2019-12-04)
Lina M González, Nikita Mukhitov, Christopher A Voigt
ABSTRACT
Materials can be made multifunctional by embedding them with living cells that perform sensing, synthesis, energy production, and physical movement. A challenge is that the conditions needed for living cells are not conducive to materials processing and require continuous water and nutrients. Here, we present a three dimensional (3D) printer that can mix material and cell streams to build 3D objects. Bacillus subtilis spores were printed within the material and germinated on its exterior surface, including spontaneously in new cracks. The material was resilient to extreme stresses, including desiccation, solvents, osmolarity, pH, ultraviolet light, and γ-radiation. Genetic engineering enabled the bacteria to respond to stimuli or produce chemicals on demand. As a demonstration, we printed custom-shaped hydrogels containing bacteria that can sense or kill Staphylococcus aureus, a causative agent of infections. This work demonstrates materials endued with living functions that can be used in applications that require storage or exposure to environmental stresses.
Sigma-Aldrich
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Sigma-Aldrich
Thiamine hydrochloride, reagent grade, ≥99% (HPLC)
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Sigma-Aldrich
Manganese(II) chloride, powder and chunks, ≥99% trace metals basis
Millipore
Peptone from soybean, enzymatic digest, suitable for microbiology
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Magnesium sulfate heptahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
4-Isopropylbenzoic acid, ≥96%
Sigma-Aldrich
Calcium nitrate hydrate, 99.997% trace metals basis
Sigma-Aldrich
Cellulase from Trichoderma reesei, aqueous solution, ≥700 units/g
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Calcium chloride, anhydrous, granular, ≤7.0 mm, ≥93.0%
Sigma-Aldrich
D-(+)-Xylose, 99%, natural sourced
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%