Merck
  • Suppressing Aneuploidy-Associated Phenotypes Improves the Fitness of Trisomy 21 Cells.
Suppressing Aneuploidy-Associated Phenotypes Improves the Fitness of Trisomy 21 Cells.
Cell reports (2019-11-21)
Sunyoung Hwang, Jessica F Williams, Maja Kneissig, Maria Lioudyno, Isabel Rivera, Pablo Helguera, Jorge Busciglio, Zuzana Storchova, Megan C King, Eduardo M Torres
ABSTRACT
An abnormal number of chromosomes, or aneuploidy, accounts for most spontaneous abortions, causes developmental defects, and is associated with aging and cancer. The molecular mechanisms by which aneuploidy disrupts cellular function remain largely unknown. Here, we show that aneuploidy disrupts the morphology of the nucleus. Mutations that increase the levels of long-chain bases suppress nuclear abnormalities of aneuploid yeast independent of karyotype identity. Quantitative lipidomics indicates that long-chain bases are integral components of the nuclear membrane in yeast. Cells isolated from patients with Down syndrome also show that abnormal nuclear morphologies and increases in long-chain bases not only suppress these abnormalities but also improve their fitness. We obtained similar results with cells isolated from patients with Patau or Edward syndrome, indicating that increases in long-chain bases improve the fitness of aneuploid cells in yeast and humans. Targeting lipid biosynthesis pathways represents an important strategy to suppress nuclear abnormalities in aneuploidy-associated diseases.
Sigma-Aldrich
D-erythro-Dihydrosphingosine, ≥98%
Sigma-Aldrich
Myriocin from Mycelia sterilia, ≥98% (HPLC), powder
Sigma-Aldrich
Phytosphingosine hydrochloride
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
N-Palmitoyl-D-sphingosine, ≥98.0% (TLC)
Sigma-Aldrich
N-Stearoyl-D-sphingosine, ≥98.0% (TLC)
Sigma-Aldrich
Anti-GAPDH Antibody, from chicken, purified by affinity chromatography
Avanti
Sphinganine (d18:0), Avanti Research - A Croda Brand
Sigma-Aldrich
(2S,3S,4R)-2-Amino-1,3,4-octadecanetriol, AldrichCPR
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial