Skip to Content
Merck
All Photos(1)

Key Documents

43799

Sigma-Aldrich

N-Palmitoyl-D-sphingosine

≥98.0% (TLC)

Synonym(s):

Cer(d18:1/16:0), (2S,3R,4E)-2-(Palmitoylamino)-4-octadecene-1,3-diol, N-Hexadecanoyl-D-sphingosine, N-Palmitoyl-D-erythro-sphingosine, N-[(1S,2R,3E)-2-Hydroxy-1-(hydroxymethyl)-3-heptadecen-1-yl]hexadecanamide, C16 Ceramide (d18:1/16:0), C16-Ceramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H67NO3
CAS Number:
Molecular Weight:
537.90
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.85

Assay

≥98.0% (TLC)

form

powder or solid

composition

carbon content, 75.92%
hydrogen content, 12.55%
nitrogen content, 2.60%

functional group

amide

lipid type

sphingolipids

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C34H67NO3/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-33(37)32(31-36)35-34(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h27,29,32-33,36-37H,3-26,28,30-31H2,1-2H3,(H,35,38)/b29-27+/t32-,33+/m0/s1

InChI key

YDNKGFDKKRUKPY-TURZORIXSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Almeida et al.
PloS one, 13(8), e0201373-e0201373 (2018-08-11)
Biological membranes contain a large variety of lipids species compartmentalized in different domains heterogeneous in size, composition and dynamics. Cholesterol induces membrane ordered domains thanks to its affinity for saturated lipids. Membrane domains had been studied with fluorescent probes either
Sharon Epstein et al.
Journal of lipid research, 53(3), 412-420 (2012-01-03)
Sphingolipids are not only important components of membranes but also have functions in protein trafficking and intracellular signaling. The LCB1 gene encodes a subunit of the serine palmitoyltransferase, which is responsible for the first step of sphingolipid synthesis. Here, we
Sherry S W Leung et al.
Biophysical journal, 103(12), 2465-2474 (2012-12-25)
Ceramide produced from sphingomyelin in the plasma membrane is purported to affect signaling through changes in the membrane's physical properties. Thermal behavior of N-palmitoyl sphingomyelin (PSM) and N-palmitoyl ceramide (PCer) mixtures in excess water has been monitored by ²H NMR
Gloria I Perez et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 19(7), 860-862 (2005-02-25)
An age-dependent acceleration of apoptosis occurs in female germ cells (oocytes), and this requires communication between the oocyte and its surrounding somatic (cumulus) cells. Here we show in aged mice that ceramide is translocated from cumulus cells into the adjacent
Daniel Dindo et al.
Molecular cancer therapeutics, 5(6), 1520-1529 (2006-07-05)
Ceramides are sphingolipid second messengers that are involved in the mediation of cell death. There is accumulating evidence that mitochondria play a central role in ceramide-derived toxicity. We designed a novel cationic long-chain ceramide [omega-pyridinium bromide D-erythro-C16-ceramide (LCL-30)] targeting negatively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service